Skip to Content
Merck
All Photos(1)

Key Documents

8.22185

Sigma-Aldrich

TWEEN® 40

for synthesis

Synonym(s):

Polyoxyethylene sorbitan monopalmitate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.22

Quality Level

form

liquid

potency

>38400 mg/kg LD50, oral (Rat)

bp

>100 °C

transition temp

flash point >149 °C

density

1.09 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C26H50O10/c1-2-3-4-5-6-7-8-9-10-11-24(30)34-19-18-31-20-22(32-15-12-27)26-25(35-17-14-29)23(21-36-26)33-16-13-28/h22-23,25-29H,2-21H2,1H3

InChI key

HMFKFHLTUCJZJO-UHFFFAOYSA-N

Analysis Note

Hydroxyl value: 89 - 105
Saponification value: 41 - 52
Acid value: ≤ 3
Density (d 20 °C/ 4 °C): 1.080 - 1.095
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service