Skip to Content
Merck
All Photos(2)

Key Documents

8.21619

Sigma-Aldrich

3-(Triethoxysilyl)propylamine

for synthesis

Synonym(s):

3-(Triethoxysilyl)propylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H23NO3Si
CAS Number:
Molecular Weight:
221.37
MDL number:
UNSPSC Code:
12352103
EC Index Number:
213-048-4
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

300 °C

potency

1780 mg/kg LD50, oral (Rat)
3800 mg/kg LD50, skin (Rabbit)

expl. lim.

0.8-4.5 % (v/v)

pH

11 (20 °C, 20 g/L in H2O)

bp

217 °C/1013 hPa

mp

<-70 °C

transition temp

flash point 93 °C

density

0.95 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C9H23NO3Si/c1-4-11-14(12-5-2,13-6-3)9-7-8-10/h4-10H2,1-3H3

InChI key

WYTZZXDRDKSJID-UHFFFAOYSA-N

Application


  • Synthesis of novel silane-based carboxylic acids: The study focuses on the synthesis of novel silane-based carboxylic acids for use in methacrylated epoxy-based oligomers. These compounds were shown to enhance the properties of UV-curable systems by reacting 3-(triethoxysilyl)propylamine with various carboxylic acids, thereby improving the performance of polycarbonate panels used in these experiments (N Ranjbar, M Kolahdoozan… - Journal of Macromolecular Science, Part B, 2023 - Taylor & Francis).

  • CO2/H2O adsorption on functionalized mesoporous silicas: This research investigated the enhancement of CO2 and water adsorption capacities on mesoporous silicas modified with 3-(triethoxysilyl) propylamine (APTES). The study demonstrates APTES′s ability to significantly improve CO2 affinity on modified silicas, offering potential improvements for environmental applications (GL Avellaneda, R Denoyel, I Beurroies - Microporous and Mesoporous Materials, 2024 - Elsevier).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.947 - 0.952
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service