Skip to Content
Merck
All Photos(2)

Key Documents

8.07484

Sigma-Aldrich

Polyethylene glycol

300, for synthesis

Synonym(s):

Poly(ethylene glycol), Polyethylene glycol 300, Polyglycol, Polyethylene oxide, Polyoxy ethylene, PEG 300, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
13111021
EC Index Number:
500-038-2
NACRES:
NA.22

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

370 °C

potency

28000 mg/kg LD50, oral (Rat)
>20000 mg/kg LD50, skin (Rabbit)

pH

4-7 (20 °C, 100 g/L in H2O)

bp

>220 °C/1013 hPa (decomposes)

mp

-15--10 °C

transition temp

flash point 220 °C

density

1.13 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • In vitro ventricular shunt material testing model: A study developed an in vitro testing model for ventricular shunt materials, highlighting the utility of Polyethylene Glycol (PEG) as an antifouling coating, potentially improving shunt performance in clinical settings (Prindeze et al., 2024).

  • Antibacterial and anti-inflammatory properties in hydrogels: Research on GelMA-PEG hydrogels incorporated with Imidazolium Poly(ionic liquids) microspheres demonstrated enhanced antibacterial and anti-inflammatory properties, suggesting applications in medical coatings and drug delivery systems (Zhou et al., 2024).

  • Bone regeneration scaffolds: A study described the development of a Gellan Gum, Polyethylene Glycol, and Hydroxyapatite composite scaffold, enhanced with Ginseng derived Compound K, which offers potential applications in bone regeneration and tissue engineering (Thangavelu et al., 2024).

  • Uniform Polyethylene Glycol synthesis: A comprehensive review discussed the advances and challenges in achieving uniform Polyethylene Glycol (PEG), which is critical for its consistency in various industrial and pharmaceutical applications (Bento et al., 2024).

  • AI-Driven optimization of electrospun scaffolds: A study utilized artificial intelligence to optimize PCL/PEG electrospun scaffolds, significantly enhancing in vivo wound healing capabilities, highlighting its potential for medical textiles and tissue engineering applications (Virijević et al., 2024).

Linkage

Replaces: PX1286A-2; PX1286A

Analysis Note

Hydroxyl value: 356 - 394
Density (d 20 °C/ 4 °C): 1.125 - 1.127
Average molecular mass: 285 - 315
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

281.5 °F - closed cup

Flash Point(C)

138.6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service