Skip to Content
Merck
All Photos(1)

Key Documents

8.02411

Sigma-Aldrich

Chloroacetyl chloride

for synthesis

Synonym(s):

Chloroacetyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
112.94
MDL number:
UNSPSC Code:
12352100
EC Index Number:
201-171-6
NACRES:
NA.22

vapor pressure

25.3 hPa ( 20 °C)

Quality Level

form

liquid

potency

120 mg/kg LD50, oral (Rat)
662 mg/kg LD50, skin (Rat)

bp

105-110 °C/1013 hPa

mp

-22.5 °C

density

1.417 g/cm3 at 20 °C

InChI

1S/C2H2Cl2O/c3-1-2(4)5/h1H2

InChI key

VGCXGMAHQTYDJK-UHFFFAOYSA-N

General description

for synthesis

Application

Chloroacetyl chloride for synthesis. CAS 79-04-9, molar mass 112.94 g/mol.

Analysis Note

Assay (GC, area%)≥ 98.0 % (a/a)Density (d 20 °C/ 4 °C)1.417 - 1.420Identity (IR)passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1A - STOT RE 1

Target Organs

Lungs

Supplementary Hazards

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service