Skip to Content
Merck
All Photos(1)

Key Documents

8.01812

Sigma-Aldrich

Benzylamine

for synthesis

Synonym(s):

Benzylamine, Phenylmethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9N
CAS Number:
Molecular Weight:
107.15
MDL number:
UNSPSC Code:
12352116
EC Index Number:
202-854-1
NACRES:
NA.22

vapor pressure

1.2 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

405 °C

potency

552 mg/kg LD50, oral (Rat)
1350 mg/kg LD50, skin (Rat)

expl. lim.

0.7-8.2 % (v/v)

pH

11.4 (20 °C, 100 g/L in H2O)

mp

-42.8 °C

transition temp

flash point 72 °C

density

0.98 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C7H9N/c8-6-7-4-2-1-3-5-7/h1-5H,6,8H2

InChI key

WGQKYBSKWIADBV-UHFFFAOYSA-N

Application

  • Organic synthesis in water with covalent organic frameworks: Research on fully conjugated covalent organic framework nanosheets, facilitated by catalysts like benzylamine, demonstrates their effectiveness in visible-light-driven organic synthesis, promoting environmentally friendly chemical processes (Ma et al., 2024).
  • Photocatalytic oxidation of amines with microporous polymers: Demonstrates the confined synthesis of conjugated microporous polymers, utilizing benzylamine as a substrate for selective photocatalytic oxidation of amines, a technique applicable in the purification of chemical substances and environmental cleanup (You et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.982 - 0.983
Water (K. F.): ≤ 0.25 %
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service