Skip to Content
Merck
All Photos(1)

Key Documents

70607

Millipore

4X SDS Sample Buffer

Synonym(s):

SDS sample buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105337
NACRES:
NC.01

form

solution

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

ambient
dry ice

storage temp.

−20°C

General description

The 4X SDS Sample Buffer is a standard formulation commonly used for SDS-PAGE analysis of proteins. The solution includes DTT for complete denaturation of disulfide bonds. The buffer can be used at 2X for most applications.

Application

4X SDS Sample Buffer has been used to denature protein lysates for western blot analysis. It has also been used to boil immunoprecipitated complex for western blot.

Warning

Toxicity: Harmful (C)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

PP2A protects podocytes against Adriamycin-induced injury and epithelial-to-mesenchymal transition via suppressing JIP4/p38-MAPK pathway
Lu Z, et al.
Cytotechnology, 1-17 (2021)
Ana Artero-Castro et al.
International journal of molecular sciences, 22(4) (2021-03-07)
Hereditary retinal dystrophies (HRD) represent a significant cause of blindness, affecting mostly retinal pigment epithelium (RPE) and photoreceptors (PRs), and currently suffer from a lack of effective treatments. Highly specialized RPE and PR cells interact mutually in the functional retina

Articles

Explore the impact of water quality on protein electrophoresis and Western blotting. Learn about the role of ultrapure water and its effect on chemiluminescent signals in Western blotting.

Explore the impact of water quality on protein electrophoresis and Western blotting. Learn about the role of ultrapure water and its effect on chemiluminescent signals in Western blotting.

Explore the impact of water quality on protein electrophoresis and Western blotting. Learn about the role of ultrapure water and its effect on chemiluminescent signals in Western blotting.

Explore the impact of water quality on protein electrophoresis and Western blotting. Learn about the role of ultrapure water and its effect on chemiluminescent signals in Western blotting.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service