Skip to Content
Merck
All Photos(1)

Key Documents

69203

Millipore

Streptavidin Agarose

Column or batch isolation of biotinylated molecules

Synonym(s):

Streptavidin beads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

Quality Level

form

liquid

manufacturer/tradename

Novagen®

storage condition

do not freeze

shipped in

ambient

storage temp.

2-8°C

General description

Specifications
Concentration50% suspension (5 ml settled volume)
Storage buffer100 mM sodium phosphate pH 7.5, 0.02% sodium azide
Size40-165 µm
Binding capacity>85 nmol free biotin/ml
ProteaseNone detected (SDS-PAGE of purified protein)
DNaseNone detected (blue/white colony assay of ligated plasmid)
RNaseNone detected (gel assay of purified RNA)
Column or batch isolation of biotinylated molecules
Streptavidin Agarose consists of cross-linked agarose covalently coupled with pure streptavidin under controlled conditions. The stable linkage to the resin minimizes leaching of the streptavidin while maintaining full binding activity. The matrix is suitable for use in column and batch formats for any application that requires high biotin binding capacity and low non-specific binding. The resin has no detectable protease, DNase, or RNase contamination. Provided as a 50% slurry in storage buffer; 5 ml settled resin in 10 ml total volume.

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huanzhou Xu et al.
PLoS pathogens, 15(12), e1008228-e1008228 (2019-12-17)
Epstein-Barr virus (EBV) is an oncogenic herpesvirus and WHO class 1 carcinogen that resides in B lymphocytes of nearly all humans. While silent in most, EBV can cause endemic Burkitt lymphoma in children and post-transplant lymphoproliferative disorders/lymphomas in immunocompromised hosts.
Chinnadurai Mani et al.
Oncogene, 39(12), 2583-2596 (2020-01-29)
Phosphofurin acidic cluster sorting protein-1 (PACS-1) is a multifunctional membrane traffic regulator that plays important roles in organ homeostasis and disease. In this study, we elucidate a novel nuclear function for PACS-1 in maintaining chromosomal integrity. PACS-1 progressively accumulates in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service