Skip to Content
Merck
All Photos(1)

Key Documents

627610

Sigma-Aldrich

Z-VAD-FMK

Single spot by TLC, solid, Caspase inhibitor, Calbiochem

Synonym(s):

Caspase Inhibitor I, Z-VAD (OMe)-FMK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30FN3O7
CAS Number:
Molecular Weight:
467.49
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Caspase Inhibitor I, Z-VAD-FMK, CAS 187389-52-2, is a cell-permeable, irreversible, pan-caspase inhibitor.

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

DMSO: 50 mM

shipped in

ambient

storage temp.

−20°C

InChI

1S/C22H30FN3O7/c1-13(2)19(26-22(31)33-12-15-8-6-5-7-9-15)21(30)24-14(3)20(29)25-16(17(27)11-23)10-18(28)32-4/h5-9,13-14,16,19H,10-12H2,1-4H3,(H,24,30)(H,25,29)(H,26,31)/t14-,16-,19-/m0/s1

InChI key

MIFGOLAMNLSLGH-QOKNQOGYSA-N

General description

A cell-permeable, irreversible, pan-caspase inhibitor. Inhibits Fas-mediated apoptosis in Jurkat cells and staurosporine-induced cell death in corneal epithelial cells. Shown to enhance the freeze-thaw survival of human embryonic stem cells. When using with purified native or recombinant enzyme, pre-treatment with an esterase is required.
A cell-permeable, irreversible, pan-caspase inhibitor. Shown to enhance the freeze-thaw survival of human embryonic stem cells. Inhibits Fas-mediated apoptosis in Jurkat T cells. Also reported to inhibit Peptide: N-glycanse (PNGase) in vitro amd in vivo. When using with a purified recombinant enzyme, pretreatment with an esterase is required.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Fas-mediated apoptosis in Jurkat cells and staurosporine-induced cell death in corneal epithelial cells
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Val-Ala-Asp(OMe)-CH₂F*

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for 6 months at -20°C.

Analysis Note

Single spot by TLC

Other Notes

Heng, B.C., et al. 2007. Biosci. Rep.27, 257.
Martin, U., et al. 2007. Apoptosis12, 525.
Misaghi, S., et al. 2004. Chem. Biol.11, 1677.
Joo, C., et al. 1999. Ophthalmic Res. 31, 287.
McColl, K.S., et al. 1998. Mol. Cell. Endocrinol. 139, 229.
Thornberry, N.A. and Lazebnik, Y. 1998. Science 281, 1312.
Tolomeo, M., et al. 1998. Cell Death Differ. 5, 735.
An, S. and Know, K.A. 1996. FEBS Lett.386, 115.
Cain, K., et al. 1996. Biochem J.314, 27.
Mirzoeva, O.K., et al. 1996. FEBS Lett.396, 266.
Pronk, G.J., et al. 1996. Science 271, 808.
Slee, E.A., et al. 1996. Biochem. J.315, 21.
Chow, S.C., et al. 1995. FEBS Lett. 364, 134.
Fearnhead, H.O., et al. 1995. FEBS Lett. 375, 283.
Zhu, H., et al. 1995. FEBS Lett.374, 303.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael J Twiner et al.
Chemical research in toxicology, 25(7), 1493-1501 (2012-06-26)
Azaspiracids (AZAs) are polyether marine dinoflagellate toxins that accumulate in shellfish and represent an emerging human health risk. Although there have been no deaths associated with the AZA toxins, humans exposed to AZAs experience severe gastrointestinal symptoms. This toxin class
A J Dowling et al.
Cellular microbiology, 6(4), 345-353 (2004-03-11)
Photorhabdus bacteria produce a number of toxins to kill their insect hosts. The expression of one of these, Makes caterpillars floppy (Mcf), is sufficient to allow Escherichia coli to persist within and kill caterpillars. Mcf causes shedding of the insect
Nam-Sook Park et al.
Journal of cellular and molecular medicine, 25(17), 8300-8311 (2021-07-29)
Dasatinib is an inhibitor of Src that has anti-tumour effects on many haematological and solid cancers. However, the anti-tumour effects of dasatinib on human oral cancers remain unclear. In this study, we investigated the effects of dasatinib on different types
Akash Saha et al.
Cell death discovery, 7(1), 356-356 (2021-11-17)
Alzheimer's disease (AD) is characterized by accumulation of senile amyloid-β (Aβ) plaques and hyperphosphorylated tau tangles causing progressive loss of synapse and neuronal death. Out of the various neuron death modalities, autophagy and apoptosis are reported to be the major
Jiyoung Lee et al.
Molecules and cells, 45(4), 193-201 (2022-03-16)
Excessive production of reactive oxygen species (ROS) is a key phenomenon in tumor necrosis factor (TNF)-α-induced cell death. However, the role of ROS in necroptosis remains mostly elusive. In this study, we show that TNF-α induces the mitochondrial accumulation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service