Skip to Content
Merck
All Photos(1)

Key Documents

573108-M

Millipore

STAT5 Inhibitor

The STAT5 Inhibitor, also referenced under CAS 285986-31-4, controls the biological activity of STAT5.

Synonym(s):

STAT5 Inhibitor, Nʹ-((4-Oxo-4H-chromen-3-yl)methylene)nicotinohydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11N3O3
CAS Number:
Molecular Weight:
293.28
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 25 mg/mL

storage temp.

2-8°C

General description

A cell-permeable nonpeptidic nicotinoyl hydrazone compound that selectively targets the SH2 domain of STAT5 (IC50 = 47 µM against STAT5b SH2 domain EPO peptide binding activity), while exhibiting much less effect towards the SH2 domain of STAT1, STAT3, or Lck (IC50 >500 µM). Shown to block STAT5/STAT5 DNA binding activity in K562 nuclear extract and inhibit IFN-α-stimulated STAT5, but not STAT1 or STAT3, tyrosine phosphorylation in Daudi cells.

Warning

Toxicity: Standard Handling (A)

Other Notes

Muller, J., et al. 2008. ChemBioChem9, 723.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service