Skip to Content
Merck
All Photos(1)

Key Documents

5.30351

Sigma-Aldrich

Nrf2 Activator IV, VSC2

Synonym(s):

Nrf2 Activator IV, VSC2, ( E)-1-Chloro-2-(2-(2-methoxyphenylsulfonyl)vinyl)benzene, Nuclear Factor E2-related Factor 2 Activator IV, VSC2, (E)-1-Chloro-2-(2-(2-methoxyphenylsulfonyl)vinyl)benzene, Nuclear Factor E2-related Factor 2 Activator IV, VSC2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13ClO3S
CAS Number:
Molecular Weight:
308.78
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable vinylsulfone compound that acts as a more potent Nrf-2 activator than the analogous α,β-unsaturated ketone-based chalcones (e.g. Cat. No. 492040) in stimulating HO-1 expression in murine microglial BV-2 cultures (382% of control protein level in 24 h; 20 µM). Drug treatment in murine dopaminergic (DAergic) CATH.a cultures likewise effectively induces Nrf2 nuclear translocation (Fold of control in 3 h/dose = 1.4/1 µM, 2.3/5 µM, 3.4/10 µM) and overall Nrf2 cellular accumulation (Fold of control in 24 h/dose = 1.5/0.5 µM, 2.0/1 µM, 2.8/2 µM, 4.3/5 µM), presumably by freeing Nrf2 from its negative regulator keap1, with concomitant upregulation of Nrf2-regulated antioxidant enzymes expression, including heme oxygenase-1/OH-1, NAD(P)H quinone oxidoreductase 1/NQO1, glutamate-cysteine ligase modifier & catalytic subunits, QCLM & GCLC. Shown to protect CATH.a against BH4 toxicity (effective conc. 0.1 to 1.0 µM against 200 µM BH4 for 24 h) in vitro and prevent MPTP- (Cat. No. 506382) induced motor deficits/dysfunctions (by vertical grid test 6 d post MPTP injection) and neurodegeneration (by TH staining 7 d post MPTP injection) in a murine PD model in vivo (3 X 10 mg drug/kg/d oral dosages; 4X 20 mg MPTP/kg/2 h i.p. dosages starting 24 h post the 1st drug dosage).
A cell-permeable vinylsulfone that acts as a more potent Nrf-2 activator than the chalcones (e.g. Cat. No. 492040) in stimulating HO-1 expression in murine microglial BV-2 cultures (3.82-fold of control in 24 h; 20 µM). Shown to induce overall Nrf2 cellular accumulation & nuclear translocation (1 to10 µM) in murine dopaminergic CATH.a cultures, presumably by freeing Nrf2 from its negative regulator keap1, with concomitant upregulation of Nrf2-regulated antioxidant enzymes expression, effectively protecting CATH.a against BH4 toxicity (effective conc. 0.1 to 1.0 µM against 200 µM BH4; 24 h) in vitro and preventing MPTP- (Cat. No. 506382) induced motor dysfunctions and neurodegeneration in a murine PD model in vivo (3X 10 mg drug/kg/d p.o.; 4X 20 mg MPTP/kg/2 h i.p.).

Biochem/physiol Actions

Cell permeable: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Use only fresh DMSO for reconstitution.

Other Notes

Lee, J.A., et al. 2015. Br. J. Pharmacol.172, 1087.
Woo, S.Y., et al. 2014. J. Med. Chem.57, 1473.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service