Skip to Content
Merck
All Photos(2)

Key Documents

516483

Sigma-Aldrich

Pepstatin A, Penetratin

A Pepstatin A that is ~75% less potent against Cathespins D/E than PepA in cell-free assays.

Synonym(s):

Pepstatin A, Penetratin, PepA-RQIKIWFQNRRMKWKK-OH, PepA-Antp₄₃₋₅₈

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C138H229N39O28S
Molecular Weight:
2914.60
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 1 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A Pepstatin A (PepA; Cat. No. 516481) penetratin (pAntp43-58) conjugate (PepA-P) that is ~75% less potent against Cathespins D/E (IC50 = 920 nM) than PepA in cell-free assays, presumably due to its decreased solubility. However, with its enhanced permeability, PepA-P is much more active than PepA in inhibiting cellular aspartic protease activity in MCF7 (94.1% vs. 54.6% inhibition wtih 10 µM respective compound), Boleths, and Dendritic cultures. PepA-P, but not PepA, effectively interferes with antigen processing by primary APCs and the subsequent activation of antigen-specific memory T cells.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

PepA-Arg-Gln-Ile-Lys-Ile-Trp-Phe-Gln-Asn-Arg-Arg-Met-Lys-Trp-Lys-Lys-CO₂H

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zaidi, N., et al. 2007. Biochem. Biophys. Res. Commun.364, 243.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrienn Skopál et al.
The Journal of biological chemistry, 298(5), 101888-101888 (2022-04-04)
Adenosine A2A receptor (A2AR)-dependent signaling in macrophages plays a key role in the regulation of inflammation. However, the processes regulating A2AR targeting to the cell surface and degradation in macrophages are incompletely understood. For example, the C-terminal domain of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service