Skip to Content
Merck
All Photos(1)

Key Documents

428026

Sigma-Aldrich

Latrunculin A

from Latrunculia magnifica, ≥85% (HPLC), liquid, actin polymerization inhibitor, Calbiochem

Synonym(s):

InSolution Latrunculin A, Latrunculia magnifica

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H31NO5S
Molecular Weight:
421.55
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Latrunculin A, Latrunculia magnifica, InSolution, ≥85%, 1 mM in DMSO, marine toxin that disrupts microfilament organization in cultured cells

Quality Level

Assay

≥85% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable marine toxin from the Red Sea sponge Latrunculia magnifica that disrupts microfilament organization in cultured cells by the formation of a 1:1 complex with monomeric G-actin (Kd = 200 nM). Also a potent inhibitor of microfilament-mediated processes in sperm, eggs, and embryos.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Inhibitor of microfilament-mediated processes in sperm, eggs, and embryos
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Physical form

A 1 mM (50 µg/119 µl) solution of Latrunculin A, Latrunculia magnifica Cat. No. 428021 in DMSO.

Reconstitution

Following initial use, aliquot and freeze (-20°C).

Other Notes

Ayscough, K.R., et al. 1997. J. Cell Biol.137, 399.
Spector, I., et al. 1989. Cell Motil. Cytoskeleton13, 127.
Coue, M., et al. 1987. FEBS Lett.213, 316.
Schatten, G., et al. 1986. Exp. Cell Res.166, 191.
Spector, I., et al. 1983. Science219, 493.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kelvin See et al.
Developmental biology, 466(1-2), 90-98 (2020-07-28)
Spatial organization of the genome in the nucleus plays a critical role in development and regulation of transcription. A genomic region that resides at the nuclear periphery is part of the chromatin layer marked with histone H3 lysine 9 dimethyl
Aniket Ghosh et al.
eLife, 9 (2020-09-10)
Ion channel complexes promote action potential initiation at the mammalian axon initial segment (AIS), and modulation of AIS size by recruitment or loss of proteins can influence neuron excitability. Although endocytosis contributes to AIS turnover, how membrane proteins traffic to
Xing Duan et al.
Nature communications, 11(1), 277-277 (2020-01-16)
Migration of meiosis-I (MI) spindle from the cell center to a sub-cortical location is a critical step for mouse oocytes to undergo asymmetric meiotic cell division. In this study, we investigate the mechanism by which formin-2 (FMN2) orchestrates the initial
Elise Lv Malavasi et al.
eLife, 10 (2021-07-10)
Voltage-gated sodium channels cluster in macromolecular complexes at nodes of Ranvier to promote rapid nerve impulse conduction in vertebrate nerves. Node assembly in peripheral nerves is thought to be initiated at heminodes at the extremities of myelinating Schwann cells, and
Antonios Chronopoulos et al.
Nature materials, 19(6), 669-678 (2020-01-08)
Extensive research over the past decades has identified integrins to be the primary transmembrane receptors that enable cells to respond to external mechanical cues. We reveal here a mechanism whereby syndecan-4 tunes cell mechanics in response to localized tension via

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service