Skip to Content
Merck
All Photos(1)

Key Documents

1.46381

Millipore

Buffered NaCl-Peptone Solution + 0.1% Tween® 80

ready-to-use, bottle volume 1000 mL , filling volume

Synonym(s):

Buffered Saline Peptone Water with 0.1% Tween, Buffered Sodium Chloride Peptone Solution with 0.1% Polysorbate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106229
NACRES:
NA.74

Agency

EP 2.6.1
JP 4.06
USP 71

Quality Level

description

For the microbiological examination of non-sterile pharmaceuticals

sterility

sterile; autoclaved

form

liquid

shelf life

12 mo.

feature

closure type screw cap with septum
ready-to-use

packaging

single packed of 1 L (in 1 L bottle with blue screw cap and 3 loci (6 bottles per box))
bottle of

technique(s)

direct inoculation: suitable
sterile filtration: suitable

bottle capacity

1000 mL

bottle volume

1000 mL , filling volume

pH

7.0

application(s)

bioburden testing
cosmetics
food and beverages
pharmaceutical

components

tween

storage temp.

2-25°C

suitability

nonselective for

General description

Buffered NaCl-Peptone solution has low peptone, along with phosphate buffer and sodium chloride that maintains the viability of micro-organisms in the solution, without supporting further growth. The polysorbate 80 also assists in the suspension of poorly wettable substances. It further serves as a neutralizing agent for disinfectants such as quaternary ammonium compounds, iodine, and parabens.

Application

Buffered NaCl-Peptone with Tween® 80 solution is suitable for the microbial examination of non-sterile pharmaceutical products. It is also used as a solution for sample preparation or as a dilution or washing solution.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service