Skip to Content
Merck
All Photos(1)

Key Documents

1.25157

Millipore

Eshmuno® CPX Resin

MiniChrom Column Eshmuno® CPX, volume 5 mL, prepacked column

Synonym(s):

MiniChrom Column Eshmuno® CPX, 5ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

ligand

(Sulfoisobutyl)

Quality Level

material

polypropylene housing

description

8 x 100mm ESHMUNO®

product line

Eshmuno®

form

prepacked column

parameter

≤8 bar pressure
500 cm/hr max. flow rate

volume

5 mL

matrix active group

Polyvinyl Ether

mean particle size

50 μm

pH

7 ( in H2O)

capacity

~120 mL, resin binding capacity

transition temp

flash point 35 °C

density

1 g/cm3

application(s)

mAb
recombinant protein
vaccine development

separation technique

cation exchange

storage temp.

no temp limit

Features and Benefits

Eshmuno® CPX chromatography resin enables:

  • Intermediate purification or polishing of monoclonal antibodies
  • High aggregate removal efficiency combined with high dynamic binding capacity
  • High loading capacity combined with high purity at lower elution volumes.
  • Wide pH and conductivity operating window
  • High flow rate versus pressure-flow behavior
  • Excellent removal of impurities
  • Robust and safe packing procedures
  • Strong chemical stability
  • Compatibility with 1 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl and storage solutions 0.1 M NaOH

Packaging

Plastic bag(s)

Legal Information

Eshmuno is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F

Flash Point(C)

35 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Learn more on mAb downstream processing, more specifically polishing chromatography and the relevant associated products.

Learn more on mAb downstream processing, more specifically polishing chromatography and the relevant associated products.

Learn more on mAb downstream processing, more specifically polishing chromatography and the relevant associated products.

Learn more on mAb downstream processing, more specifically polishing chromatography and the relevant associated products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service