Skip to Content
Merck
All Photos(1)

Key Documents

12-372

Sigma-Aldrich

Histone H4 Peptide

biotin conjugate, residues 2-24

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

product name

Histone H4 Peptide, biotin conjugate, residues 2-24, Histone H4 Peptide, biotin conjugate, residues 2-24 primarily used in Immunoprecipitation and Affinity Precipitation Assays.

Quality Level

manufacturer/tradename

Upstate®

technique(s)

affinity precipitation assay: suitable
immunoprecipitation (IP): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Quality

Routinely evaluated by HAT Assay as a substrate in histone acetyl transferase (HAT) and deacetylase assays. For additional information, please refer to HAT and HDAC assay kits (catalog #17-289 and #17-320).

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K G Tanner et al.
The Journal of biological chemistry, 274(26), 18157-18160 (1999-06-22)
Within chromatin, reversible acetylation of core histones is critical for transcriptional activation of eukaryotic target genes. The recent identification of intrinsic histone acetyltransferase (HAT) catalytic activity from a number of transcriptional co-activators (including yeast GCN5, p300/CBP, P/CAF, and TAFII250), has
Neonatal exposure to sucralose does not alter biochemical markers of neuronal development or adult behavior.
Henrik Viberg,Anders Fredriksson
Nutrition null
S Ait-Si-Ali et al.
Nucleic acids research, 26(16), 3869-3870 (1998-08-01)
Histone acetyl-transferases (HATs) seem to be key elements in the regulation of transcription. We have designed an enzymatic assay to quantify HAT enzymatic activity. In this assay, the substrate is a peptide corresponding to the 24 first amino acids of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service