Skip to Content
Merck
All Photos(1)

Key Documents

1.10307

Supelco

Protein Kit, photometric

photometric, 0.5-10 g/L (Protein), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.21

product name

Protein Kit, photometric, 0.5-10 g/L (Protein), Spectroquant®

Quality Level

usage

sufficient for 250 tests

analyte chemical class(es)

proteins

measuring range

0.5-10 g/L (Protein)

technique(s)

photometry: suitable

pH

13.2 (20 °C in H2O)

density

1.02 g/cm3 at 20 °C

compatibility

for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Biuret Method)

storage temp.

2-8°C

General description

Biuret Method, reagent solution for approx. 250 determinations Proteins form a blue-violet complex in alkaline copper sulfate solution containing tartrate (Biuret reagent). The absorbance of the solution is normally measured at 546 nm. The protein concentration of the sample can be determined by calibrating with a protein standard solution.

Application


  • Success in treating wounds with local boric acid: a case study: Details a clinical case study where boric acid was successfully used to treat chronic wounds, showcasing its potential in medical applications (Coskun M, 2023).

  • Mechanical Force-Induced Color-Variable Luminescence of Carbon Dots in Boric Acid Matrix: Demonstrates the use of boric acid matrices to develop pressure-sensitive luminescent materials with potential applications in sensing and display technologies (Meng S et al., 2023).

  • Insight into boron-doped biochar as efficient metal-free catalyst for peroxymonosulfate activation: Investigates boric acid′s role in enhancing the catalytic properties of boron-doped biochar for environmental cleanup and pollutant degradation (Pan G et al., 2023).



Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service