Skip to Content
Merck
All Photos(2)

Key Documents

1.10001

Supelco

Peracetic Acid Test Strips, colorimetric

greener alternative

100-500 mg/L (peracetic acid), MQuant®

Synonym(s):

Peracetic Acid chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Peracetic Acid Test, colorimetric, 100-500 mg/L (peracetic acid), MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

peracetic acid

reaction suitability

reaction type: Redox Reactions

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

measuring range

100-500 mg/L (peracetic acid)

detection method

colorimetric

greener alternative category

storage temp.

2-8°C

General description

Method: Colorimetric, with test strips, 100 - 150 - 200 - 250 - 300 - 400 - 500 mg/L Peracetic acid, MQuant®

Peracetic acid is a frequently used disinfectant. Due to its measuring range the MQuant® Peracetic acid test strips are especially suitable to check the presence of effective levels in peracetic acid based disinfectants. As peracetic acid solutions are very unstable, frequent controls are necessary. Peracetic acid reacts with an aromatic amine to form a blue dye. The concentration of peracetic acid is measured semi-quantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the peracetic acid concentration and a possible dilution factor prior to using a quantitative method such as a Reflectoquant® test.

Design for sustainability benefits:

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.

This product has benefited from:

  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet

Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Application


  • Efficient activation of peracetic acid by cobalt modified nitrogen-doped carbon nanotubes for drugs degradation: This study explores a novel method to enhance the efficacy of peracetic acid in degrading pharmaceutical compounds, highlighting its potential in environmental disinfection monitoring and safety applications in pharmaceutical manufacturing (Yao et al., 2024).

  • Ecotoxicological evaluation of effluent from bovine slaughterhouses disinfected by peracetic acid (PAA): This research assesses the environmental impact of peracetic acid used in the disinfection of wastewater from bovine slaughterhouses, using the bioindicator Girardia tigrina, which is crucial for environmental safety assessments (Pereira et al., 2024).

  • Peracetic acid activated by nickel cobaltite as effective oxidizing agent for BPA and its analogues degradation: This article presents a method for activating peracetic acid with nickel cobaltite to degrade BPA and its analogs effectively, addressing concerns over biocide efficacy and environmental safety (Kiejza et al., 2024).

  • Accelerated removal of naproxen in the iron-based peracetic acid activation system by chloride ions: The study focuses on enhancing reactive oxidative species in water treatment processes, showing how chloride ions can boost peracetic acid′s efficiency, important for biocide applications and safety testing (Yu et al., 2024).

Linkage

Replaces: 10001-1; 10001

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

See All

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service