Skip to Content
Merck
All Photos(1)

Key Documents

1.08218

Supelco

Trifluoroacetic acid solution

(25% solution in water) for protein sequenation

Synonym(s):

Trifluoroacetic acid solution, TFA solution, TFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COOH
CAS Number:
Molecular Weight:
114.02
Beilstein:
5446934
MDL number:
UNSPSC Code:
12352106
NACRES:
NB.21

Quality Level

form

liquid

composition

Water, 74.5-75.5% Karl Fischer

concentration

24.5-25.5% (trifluoroacetic acid)

density

1.12 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

OC(=O)C(F)(F)F

InChI

1S/C2HF3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

DTQVDTLACAAQTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trifluoroacetic acid (TFA) can be used as a solvent in a reverse-phase HPLC system to separate peptides because it enhances peak resolution. It also has low absorption detection wavelengths and is relatively volatile and easily removed. For the majority of peptide separations, TFA has been used at concentrations ranging from 0.05 to 0.1%(6.5-13 mM).

Analysis Note

Assay (alkalimetric): 24.5 - 25.5 %
Identity (IR-spectrum): passes test
Density (d 20 °C/ 4 °C): 1.129 - 1.131
Fluorescence (at 254 nm): ≤ 2 ppb
Fluorescence (at 365 nm): ≤ 2 ppb
Water (according to Karl Fischer): 74.5 - 75.5 %

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Chen et al.
Journal of chromatography. A, 1043(1), 9-18 (2004-08-20)
Trifluoroacetic acid (TFA) remains the dominant mobile phase additive for reversed-phase high-performance liquid chromatography (RP-HPLC) of peptides after more than two decades since its introduction to this field. Generally, TFA has been employed in a concentration range of 0.05-0.1% (6.5-13

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service