Skip to Content
Merck
All Photos(1)

Key Documents

1.03649

Supelco

Ethyl acetate

hypergrade for LC-MS LiChrosolv®

Synonym(s):

Ethyl ethanoate, Acetic ester, Ethyl ester, EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
MDL number:
UNSPSC Code:
41116105
EC Index Number:
205-500-4
NACRES:
NA.03

vapor density

3 (20 °C, vs air)

Quality Level

vapor pressure

73 mmHg ( 20 °C)

product line

LiChrosolv®

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

801 °F

expl. lim.

2.2-11.5 %, 38 °F

technique(s)

LC/MS: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.05% Water
≤10 ppb Al (Aluminium)
≤10 ppb Ca (Calcium)
≤10 ppb Fe (Iron)
≤10 ppb Mg (Magnesium)
≤5 ppb K (Potassium)
≤50 ppb Na (Sodium)

evapn. residue

≤2.0 mg/L

transmittance

260 nm, ≥50%
265 nm, ≥80%
270 nm, ≥98%

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

transition temp

flash point -4 °C

density

0.902 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is a high purity solvent that ensures baseline stability, lowest impurity levels and high UV transmittance in LC-MS applications.

Application

Intended for HPLC or LC-MS, either as an extraction solvent or as a mobile phase.

Features and Benefits

  • no ghost peaks
  • full reproducibility
  • extends the lifetime of columns
  • optimized and tested for LC-MS applications
  • minimal background ion suppression
  • reduced risk of contamination
  • less exposure to hazardous chemicals

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Purity (GC): ≥ 99.8 %
Identity (IR): conforms
Evaporation residue: ≤ 2.0 mg/l
Water: ≤ 0.05 %
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Al (Aluminium): ≤ 10 ppb
Ca (Calcium): ≤ 10 ppb
Fe (Iron): ≤ 10 ppb
K (Potassium): ≤ 5 ppb
Mg (Magnesium): ≤ 10 ppb
Na (Sodium): ≤ 50 ppb
Transmission (at 260 nm): ≥ 50 %
Transmission (at 265 nm): ≥ 80 %
Transmission (from 270 nm): ≥ 98 %
Suitable for LC-MS; Intensity of background mass peak based on reserpine(APCI/ESI positive): ≤ 2 ppb
Suitable for LC-MS; Intensity of background mass peak based on reserpine(APCI/ESI negative): ≤ 20 ppb
Filtered by 0.2 μm filter.

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F

Flash Point(C)

-4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service