Skip to Content
Merck
All Photos(3)

Key Documents

1.02442

Supelco

Chloroform

for analysis (for determinations with dithizone)

Synonym(s):

TCM, Trichloromethane, Methane trichloride, Methyl trichloride, Methylidyne trichloride, Trichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
Beilstein:
1731042
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-663-8
NACRES:
NA.21

vapor density

4.1 (vs air)

Quality Level

vapor pressure

160 mmHg ( 20 °C)

Assay

99.0-99.4% (GC)

form

liquid

potency

695 mg/kg LD50, oral (Rat)
>3980 mg/kg LD50, skin (Rabbit)

impurities

≤0.0001% Chloride (Cl)
≤0.001% Free acid (as HCl)
≤0.005% Carbonyl compounds (as CO)
≤0.01% Carbon tetrachloride (GC)
≤0.01% Dichloromethane (GC)
≤0.01% Tetrachloroethylene (GC)
≤0.01% Trichloroethylene (GC)
≤0.01% Water
0.6-1% Ethanol (GC)
0.6-1.0% Ethanol (GC)

evapn. residue

≤0.001%

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

density

1.492 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.00005%
B: ≤0.000002%
Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%

storage temp.

2-30°C

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Characterization of Natural Products: Chloroform is extensively used in the extraction and isolation of natural products. A study on the characterization and pesticidal evaluation of diterpenes isolated from chloroform extracts of Hedychium spicatum rhizomes highlights its effectiveness. This application is crucial for identifying bioactive compounds and assessing their potential uses in pharmaceuticals and agrochemicals (Rawat et al., Analytical Chemistry Letters, 2023).

  • Self-Diffusion and Molecular Association Studies: Chloroform is used in studies of self-diffusion and molecular association in binary systems, such as dimethyl sulfoxide (DMSO) and acetone. These studies provide insights into the molecular interactions and dynamics in mixed solvents, which are essential for understanding solvation and reaction mechanisms in various chemical processes (Gurina and Golubev, Results in Chemistry, 2022).

  • Cell-Free DNA Isolation: A novel phenol-chloroform-free method for isolating cell-free DNA from cell-conditioned media has been developed, optimized, and compared with traditional methods. This advancement is significant for genomic studies, allowing for more efficient and less hazardous extraction of nucleic acids from biological samples (Das et al., Analytical Biochemistry, 2024).

  • Green Chemistry and Circular Economy: Chloroform plays a role in green analytical chemistry, particularly in the development of circular chemistry principles. Research emphasizes the need to design chemical processes that minimize waste and environmental impact, promoting sustainability in analytical laboratories. This includes the use of chloroform in recyclable and reusable systems to reduce solvent waste (Psillakis and Pena-Pereira, TrAC Trends in Analytical Chemistry, 2024).

Analysis Note

Purity (GC): 99.0 - 99.4 %
Identity (IR): conforms
Free acid (as HCl): ≤ 0.001 %
Chloride (Cl): ≤ 0.0001 %
Free chlorine (Cl): conforms
Carbonyl compounds (as CO): ≤ 0.005 %
Matter discolored by H₂SO₄: ≤ 10 Hazen
Ethanol (GC): 0.6 - 1.0 %
Dichloromethane (GC): ≤ 0.01 %
Carbon tetrachloride (GC): ≤ 0.01 %
Tetrachloroethylene (GC): ≤ 0.01 %
Trichloroethylene (GC): ≤ 0.01 %
Evaporation residue: ≤ 0.001 %
Water: ≤ 0.01 %
Suitability for determination with dithizone: conforms
Al (Aluminium): ≤ 0.00005 %
B (Boron): ≤ 0.000002 %
Ba (Barium): ≤ 0.00001 %
Ca (Calcium): ≤ 0.00005 %
Cd (Cadmium): ≤ 0.000005 %
Co (Cobalt): ≤ 0.000002 %
Cr (Chromium): ≤ 0.000002 %
Cu (Copper): ≤ 0.000002 %
Fe (Iron): ≤ 0.00001 %
Mg (Magnesium): ≤ 0.00001 %
Mn (Manganese): ≤ 0.000002 %
Ni (Nickel): ≤ 0.000002 %
Pb (Lead): ≤ 0.00001 %
Sn (Tin): ≤ 0.00001 %
Zn (Zinc): ≤ 0.00001 %
Stabilized with 0.6 - 1.0% ethanol.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service