Skip to Content
Merck
All Photos(2)

Key Documents

1.00452

Supelco

Nitric acid

65%, for analysis (max. 0.005ppm Hg) EMSURE® Reag. Ph Eur,ISO

Synonym(s):

Nitric acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01
MDL number:
UNSPSC Code:
12352301
NACRES:
NA.21

product name

Nitric acid 65%, for analysis (max. 0.005ppm Hg) EMSURE® Reag. Ph Eur,ISO

Agency

reag. ISO
reag. Ph. Eur.

Quality Level

vapor pressure

9.4 hPa ( 20 °C)

product line

EMSURE®

Assay

≥65.0% (alkalimetric)

form

liquid

impurities

≤0.005 ppm Hg (Mercury)
≤0.01 ppm Ag (Silver)
≤0.01 ppm As (Arsenic)
≤0.01 ppm Ba (Barium)
≤0.01 ppm Be (Beryllium)
≤0.01 ppm Cd (Cadmium)
≤0.01 ppm Co (Cobalt)
≤0.01 ppm Cu (Copper)
≤0.01 ppm Li (Lithium)
≤0.01 ppm Mn (Manganese)
≤0.01 ppm Mo (Molybdenum)
≤0.01 ppm Pb (Lead)
≤0.01 ppm Sr (Strontium)
≤0.01 ppm V (Vanadium)
≤0.02 ppm Cr (Chromium)
≤0.02 ppm Ge (Germanium)
≤0.02 ppm In (Indium)
≤0.02 ppm Ni (Nickel)
≤0.02 ppm Ti (Titanium)
≤0.02 ppm Tl (Thallium)
≤0.02 ppm Zn (Zinc)
≤0.02 ppm Zr (Zirconium)
≤0.05 ppm Al (Aluminium)
≤0.05 ppm Au (Gold)
≤0.05 ppm Ga (Gallium)
≤0.05 ppm Mg (Magnesium)
≤0.1 ppm Bi (Bismuth)
≤0.1 ppm Ca (Calcium)
≤0.1 ppm Fe (Iron)
≤0.1 ppm K (Potassium)
≤0.1 ppm Pt (Platinum)
≤0.2 ppm Chloride (Cl)
≤0.2 ppm Heavy metals (as Pb)
≤0.2 ppm Na (Sodium)
≤0.2 ppm Phosphate (PO4)
≤0.5 ppm Sulfate (SO4)
≤1 ppm Fluoride (F)

SMILES string

O[N+]([O-])=O

ign. residue

≤3 ppm (as SO4)

pH

<1 (20 °C in H2O, strongly acid)

bp

121 °C/1013 hPa

mp

-32 °C

density

1.39 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)

InChI key

GRYLNZFGIOXLOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (alkalimetric): ≥ 65.0 %
Identity: passes test
Appearance: passes test
Density (d 20/20): 1.384 - 1.416
Chloride (Cl): ≤ 0.2 ppm
Fluoride (F): ≤ 1 ppm
Phosphate (PO₄): ≤ 0.2 ppm
Sulfate (SO₄): ≤ 0.5 ppm
Heavy metals (as Pb): ≤ 0.2 ppm
Ag (Silver): ≤ 0.01 ppm
Al (Aluminium): ≤ 0.05 ppm
As (Arsenic): ≤ 0.01 ppm
Au (Gold): ≤ 0.05 ppm
Ba (Barium): ≤ 0.01 ppm
Be (Beryllium): ≤ 0.01 ppm
Bi (Bismuth): ≤ 0.1 ppm
Ca (Calcium): ≤ 0.1 ppm
Cd (Cadmium): ≤ 0.01 ppm
Co (Cobalt): ≤ 0.01 ppm
Cr (Chromium): ≤ 0.02 ppm
Cu (Copper): ≤ 0.01 ppm
Fe (Iron): ≤ 0.1 ppm
Ga (Gallium): ≤ 0.05 ppm
Ge (Germanium): ≤ 0.02 ppm
Hg (Mercury): ≤ 0.005 ppm
In (Indium): ≤ 0.02 ppm
K (Potassium): ≤ 0.1 ppm
Li (Lithium): ≤ 0.01 ppm
Mg (Magnesium): ≤ 0.05 ppm
Mn (Manganese): ≤ 0.01 ppm
Mo (Molybdenum): ≤ 0.01 ppm
Na (Sodium): ≤ 0.2 ppm
Ni (Nickel): ≤ 0.02 ppm
Pb (Lead): ≤ 0.01 ppm
Pt (Platinum): ≤ 0.1 ppm
Sr (Strontium): ≤ 0.01 ppm
Ti (Titanium): ≤ 0.02 ppm
Tl (Thallium): ≤ 0.02 ppm
V (Vanadium): ≤ 0.01 ppm
Zn (Zinc): ≤ 0.02 ppm
Zr (Zirconium): ≤ 0.02 ppm
Residue on ignition (as SO₄): ≤ 3 ppm
ISO-reagent
Minimum shelf life: see product label

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service