Skip to Content
Merck
All Photos(3)

Key Documents

07-1341

Sigma-Aldrich

Anti-PHLPP1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

PH domain and leucine rich repeat protein phosphatase, PH domain leucine-rich repeat protein phosphatase, Pleckstrin homology domain-containing family E protein 1, SCN circadian oscillatory protein, Suprachiasmatic nucleus circadian oscillatory protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

conjugate

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

purified by

affinity chromatography

species reactivity

rat, mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

immunogen sequence

KLH-conjugated linear peptide corresponding to the C-terminus of PHLPP1.

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... PHLPP1(23239)

General description

PHLPP1 is the protein phosphatase responsible for dephosphorylation of the Akt (Ser473) site. Akt (Ser473) is the TORC2 (mTOR) phosphorylation activation site.

Specificity

This antibody recognizes the C-terminal region of PHLPP1.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of PHLPP1.

Application

Anti-PHLPP1 Antibody detects level of PHLPP1 & has been published & validated for use in WB & IC.
Research Category
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling

Quality

Evaluated by Western Blot in Human brain tissue lysate.
Western Blot Analysis: A 1:1,000 dilution of this antibody detected PHLPP1 in Human brain tissue lysate.

Target description

134 kDa Observed at 110 kDa

Physical form

Affinity purified
Purified rabbit serum in buffer containing 0.1 M Tris-glycine, pH 7.4, 150 mM NaCl and 0.05% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C for 1 year from date of receipt.

Analysis Note

Control
Human brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saida Abdelli et al.
PloS one, 7(5), e35997-e35997 (2012-05-09)
We have recently shown that silencing of the brain/islet specific c-Jun N-terminal Kinase3 (JNK3) isoform enhances both basal and cytokine-induced beta-cell apoptosis, whereas silencing of JNK1 or JNK2 has opposite effects. While it is known that JNK1 or JNK2 may
Clara M Castillejo Becerra et al.
Journal of cellular biochemistry, 119(9), 7470-7478 (2018-05-19)
The protein phosphatase Phlpp1 is an essential enzyme for proper chondrocyte function. Altered Phlpp1 levels are associated with cancer and degenerative diseases such as osteoarthritis. While much is known about the post-transcriptional mechanisms controlling Phlpp1 levels, transcriptional regulation of the
Georgia Balsevich et al.
Nature communications, 8(1), 1725-1725 (2017-11-25)
The co-chaperone FKBP5 is a stress-responsive protein-regulating stress reactivity, and its genetic variants are associated with T2D related traits and other stress-related disorders. Here we show that FKBP51 plays a role in energy and glucose homeostasis. Fkbp5 knockout (51KO) mice
Medha Sharma et al.
Cell communication and signaling : CCS, 20(1), 179-179 (2022-11-16)
The aim of the present study was to determine the role of individual PHLPP isoforms in insulin signaling and insulin resistance in neuronal cells. PHLPP isoforms were either silenced or overexpressed individually, and the effects were observed on individual Akt
Changli Zhang et al.
JOR spine, 7(1), e1306-e1306 (2024-01-15)
Intervertebral disc (IVD) degeneration is associated with chronic back pain. We previously demonstrated that the phosphatase pleckstrin homology domain and leucine-rich repeat protein phosphatase (PHLPP) 1 was positively correlated with IVD degeneration and its deficiency decelerated IVD degeneration in both

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service