Skip to Content
Merck
All Photos(1)

Key Documents

05-939

Sigma-Aldrich

Anti-LSD1/BHC110 Antibody

ascites fluid, Upstate®

Synonym(s):

BRAF35-HDAC complex protein BHC110, FAD-binding protein BRAF35-HDAC complex, 110 kDa subunit, Flavin-containing amine oxidase domain-containing protein 2, amine oxidase (flavin containing) domain 2, lysine (K)-specific demethylase 1, lysine-specific hist

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

monoclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KDM1A(23028)

General description

Histone demethylase that specifically demethylates ′Lys-4′ of histone H3, a specific tag for epigenetic transcriptional activation, thereby acting as a corepressor. Acts by oxidizing the substrate by FAD to generate the corresponding imine that is subsequently hydrolyzed. Demethylates both mono- and tri-methylted ′Lys-4′ of histone H3. May play a role in the repression of neuronal genes. Alone, it is unable to demethylate H3 ′Lys-4′ on nucleosomes and requires the presence of RCOR1/CoREST to achieve such activity. May also demethylate ′Lys-9′ of histone H3, a specific tag for epigenetic transcriptional repression, thereby leading to derepression of androgen receptor target genes.

Specificity

Recognizes LSD1, Mr 110 kDa.

Immunogen

GST fusion protein corresponding to residues 23-138 of human LSD1.

Application

Anti-LSD1/BHC110 Antibody is a Mouse Monoclonal Antibody for detection of LSD1/BHC110 also known as BRAF35-HDAC complex protein BHC110, lysine (K)-specific demethylase 1 & has been validated in ChIP, IF, IHC, IP & WB.
Chromatin Immunoprecipitation:
This antibody has been reported by an independent laboratory to immunoprecipitate LSD1 from chromatin.

Immunoprecipitation:
Recommended.

Immunohistochemistry:
This antibody has been reported by an independent laboratory to detect LSD1 using paraffin-embedded tissues. (Metzger, E., 2005)

Immunofluorescence:
Recommended.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Chromatin Biology

Quality

Routinely evaluated by western blot.

Western Blot Analysis:
1:500-1:2000 dilution of this lot detected LSD1 in RIPA lysates from HeLa cells.

Target description

110 kDa

Linkage

Replaces: CBL770

Physical form

Mouse monoclonal in buffer containing 0.05% sodium azide and 30% glycerol.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations:
Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variabillity in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
HeLa cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dominika Žagar et al.
Journal of applied microbiology, 132(6), 4517-4530 (2022-03-11)
Childcare facilities act as microenvironments that facilitate and promote the selection, spread and transmission of antibiotic-resistant micro-organisms in the community. We focused on the study of antimicrobial resistance and genetic predispositions for β-lactamase production in bacterial isolates from nursery teachers'
Mohamed-Ali Hakimi et al.
The Journal of biological chemistry, 278(9), 7234-7239 (2002-12-21)
Eukaryotic genes are under the control of regulatory complexes acting through chromatin structure to control gene expression. Here we report the identification of a family of multiprotein corepressor complexes that function through modifying chromatin structure to keep genes silent. The
Lysine-specific histone demethylase 1 inhibitors control breast cancer proliferation in ER?-dependent and -independent manners.
Pollock, JA; Larrea, MD; Jasper, JS; McDonnell, DP; McCafferty, DG
ACS chemical biology null
Xing Zeng et al.
Genes & development, 30(16), 1822-1836 (2016-08-28)
Brown adipocytes display phenotypic plasticity, as they can switch between the active states of fatty acid oxidation and energy dissipation versus a more dormant state. Cold exposure or β-adrenergic stimulation favors the active thermogenic state, whereas sympathetic denervation or glucocorticoid
Daniella Brasacchio et al.
Diabetes, 58(5), 1229-1236 (2009-02-12)
Results from the Diabetes Control Complications Trial (DCCT) and the subsequent Epidemiology of Diabetes Interventions and Complications (EDIC) Study and more recently from the U.K. Prospective Diabetes Study (UKPDS) have revealed that the deleterious end-organ effects that occurred in both

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service