Skip to Content
Merck
All Photos(1)

Key Documents

05-666

Sigma-Aldrich

Anti-BRCA2 Antibody, clone 5.23

clone 5.23, Upstate®, from mouse

Synonym(s):

Anti BRCC2, Anti-BROVCA2, Anti-FACD, Anti-FAD, Anti-FAD1, Anti-FANCD, Anti-FANCD1, Anti-GLM3, Anti-PNCA2, Anti-XRCC11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5.23, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BRCA2(675)

Specificity

BRCA2

Immunogen

GST fusion protein to residues 2959-3418 of human BRCA2 protein

Application

Detect BRCA2 with Anti-BRCA2 Antibody, clone 5.23 (Mouse Monoclonal Antibody), that has been shown to work in WB, ICC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

routinely evaluated by immunoblot on in HeLa nuclear extract

Target description

390kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, and 0.05% sodium azide.
Format: Purified
Protein G Chromatography

Storage and Stability

Store at 2-8°C. Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at 2-8°C.

Analysis Note

Control
Positive Antigen Control: Catalog #12-309, Hela cell nuclear extract. Add an equal volume of Laemmli reducing sample buffer to 10 μL of extract and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced extract per lane for minigels.

Other Notes

Due to license agreement restrictions, this product cannot be purchased for resale.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loredana Moro et al.
The Journal of biological chemistry, 280(23), 22482-22491 (2005-04-05)
BRCA2 is a tumor suppressor gene that when mutated confers an increased susceptibility to developing breast and prostate carcinoma. Besides its role in mediating DNA repair, new evidence suggests that BRCA2 may also play a role in suppressing cancer cell
Jimmy M-M Kwok et al.
Molecular cancer research : MCR, 8(1), 24-34 (2010-01-14)
The transcription factor Forkhead box M1 (FOXM1) is a key regulator of cell proliferation and is overexpressed in many forms of primary cancers, leading to uncontrolled cell division and genomic instability. To address the role of FOXM1 in chemoresistance, we
Loredana Moro et al.
The Journal of biological chemistry, 281(31), 22100-22107 (2006-06-07)
Aberrant interaction of carcinoma cells with basement membranes (BM) is a fundamental pathophysiological process that initiates a series of events resulting in cancer cell invasion and metastasis. In this report, we describe the results of our investigations pertaining to the
Yongjun Tan et al.
Molecular and cellular biology, 27(3), 1007-1016 (2006-11-15)
The forkhead box M1 (FoxM1) transcription factor regulates expression of cell cycle genes essential for DNA replication and mitosis during organ repair and cancer progression. Here, we demonstrate that FoxM1-deficient (-/-) mouse embryonic fibroblasts and osteosarcoma U2OS cells depleted in
Wen-Yuan Wang et al.
Nature neuroscience, 16(10), 1383-1391 (2013-09-17)
Defects in DNA repair have been extensively linked to neurodegenerative diseases, but the exact mechanisms remain poorly understood. We found that FUS, an RNA/DNA-binding protein that has been linked to amyotrophic lateral sclerosis (ALS) and frontotemporal lobar degeneration, is important

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service