Skip to Content
Merck
All Photos(1)

Key Documents

05-443

Sigma-Aldrich

Anti-VEGF Antibody, clone JH

clone JH, Upstate®, from mouse

Synonym(s):

Vascular permeability factor, vascular endothelial growth factor, vascular endothelial growth factor A, vascular endothelial growth factor isoform VEGF165

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

100
400

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

JH, monoclonal

species reactivity

rat, mouse, rabbit, human

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable (paraffin)

isotype

IgG1κ

suitability

not suitable for Western blot

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... VEGFA(7422)

General description

The Flt gene family of receptor tyrosine kinases includes Flt-1, KDR/Flk-1, Flt-3, and Flt-4. They encode receptors for VEGF, characterized by seven Ig-loops within their extracellular domain and a split kinase domain within the cytoplasmic domain. VEGF is an important angiogenic factor of many solid tumors and is also involved in the differentiation of endothelial cells. Flt-1 (Fms-like tyrosine kinase, VEGF-R1) is expressed on endothelial cells and on the common precursors of endothelial and hematopoietic stem cells. KDR (Flk-1, VEGF-R2) is the principal receptor involved in VEGF signaling. KDR is expressed on endothelial cells and stimulates angiogenesis, but is also expressed on bone marrow cells, and is thought to play a role in driving the proliferation of most or all leukemic cells. Flt3 is a class III receptor tyrosine kinase that is closely related to c-Kit and c-Fms. It is expressed in primitive hematopoietic cells and its ligand exerts proliferative effects on these cells in co-operation with other cytokines. Flt3 is also expressed the placenta, testis, spleen, thymus, bone marrow, brain and in cerebellar Purkinje cells. The VEGF receptors stimulate the Ras-MAP kinase pathway, suggesting that it signals as a conventional receptor tyrosine kinase.

Specificity

Other species not tested.
Recognizes 34-54 kDa isoforms of Vascular Endothelial Growth Factor/Vascular Permeability Factor (VEGF/VPF).

Immunogen

Recombinant human VEGF121. Clone JH121

Application

Immunohistochemistry: This lot of antibody detected VEGF in formalin fixed, paraffin embedded, human.tissues, using a 1:50 dilution incubated overnight at 4°C.

Immunoprecipitation: 2 µg of this antibody has been reported to immunoprecipitate VEGF from 1 mg of protein lysate.
Research Category
Signaling
Research Sub Category
Growth Factors & Receptors
This Anti-VEGF Antibody, clone JH is validated for use in IH(P) for the detection of VEGF.

Quality

Biological Blockade: Incubation of HUVEC cells with 5 µg/mL of a previous lot and 5 ng/mL of VEGF (Catalog # 01-185) has shown growth inhibition of 20-70%.

Target description

34-54 kDa isoforms

Physical form

Format: Purified
Protein G Chromatography
Purified mouse monoclonal IgG1k in buffer containing 10 mM PBS, pH 7.4.

Storage and Stability

Stable for 1 year at from date of receipt.

Analysis Note

Control
Expressed in most tissue, including Pancreas, Kidney, Adenocarcinoma.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Exogenous Smad3 accelerates wound healing in a rabbit dermal ulcer model.
Sumiyoshi, K; Nakao, A; Setoguchi, Y; Okumura, K; Ogawa, H
The Journal of Investigative Dermatology null
Inducement of mitogen-activated protein kinases in frozen shoulders.
Kanbe, K; Inoue, K; Inoue, Y; Chen, Q
Journal of Orthopaedic Science : Official Journal of the Japanese Orthopaedic Association null
Gökhan Akarca et al.
Journal of food science and technology, 56(12), 5253-5261 (2019-11-22)
Spices are primarily used as flavor enhancers and have attracted attention as natural food preservatives since their antimicrobial effects were determined. In the present study, the antimicrobial effects, minimum inhibitory concentration (MIC) and minimum bactericidal concentration (MBC) values on 5
M Wartenberg et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 15(6), 995-1005 (2001-04-09)
Tumor vascularization is the rate-limiting step for the progression of cancer. Differential steps of tumor-induced angiogenesis were studied by a novel in vitro confrontation culture of avascular multicellular prostate tumor spheroids and embryoid bodies grown from pluripotent embryonic stem (ES)
C O Tibesku et al.
The open orthopaedics journal, 5, 177-180 (2011-06-03)
To evaluate the expression of VEGF by chondrocytes of hyaline cartilage during the course of osteoarthritis (OA). In 12 white New Zealand rabbits the anterior cruciate ligament (ACL) was resected to create an anterior instability of the knee. In 12

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service