Skip to Content
Merck
All Photos(1)

Key Documents

T-050

Supelco

Temazepam glucuronide lithium salt solution

100 μg/mL in methanol (as free carboxylate), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Temazepam Glucuronide Solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20ClLiN2O8
Molecular Weight:
482.80
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland)

concentration

100 μg/mL in methanol (as free carboxylate)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[Li+].CN1C(=O)C(O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C([O-])=O)N=C(c3ccccc3)c4cc(Cl)ccc14

InChI

1S/C22H21ClN2O8.Li/c1-25-13-8-7-11(23)9-12(13)14(10-5-3-2-4-6-10)24-19(20(25)29)33-22-17(28)15(26)16(27)18(32-22)21(30)31;/h2-9,15-19,22,26-28H,1H3,(H,30,31);/q;+1/p-1/t15-,16-,17+,18-,19?,22-;/m0./s1

InChI key

RQHXRNLYSZXQKI-NMJPLRNWSA-M

General description

The major urinary and blood metabolite of temazepam, an intermediate-acting 3-hydroxy benzodiazepine prescribed for the short-term treatment of sleeplessness in patients with difficulties remaining asleep. This new Certified Reference Standard is suitable for a variety of LCMS and GCMS applications, including forensic and forensic toxicology analysis, urine drug testing, and hydrolysis controls.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G W Kunsman et al.
Journal of chromatography, 568(2), 427-436 (1991-08-23)
A rapid and sensitive method for extracting temazepam from human serum and urine is presented. Free temazepam is extracted from plasma and urine samples using n-butyl chloride with nitrazepam as the internal standard. Temazepam glucuronide is analyzed as free temazepam
Commentary on: Dou C, Bournique J, Zinda M, Gnezda M, Nally A, Salamone S. Comparison of rates of hydrolysis of lorazepam-glucuronide, oxazepam-glucuronide and temazepam-glucuronide catalyzed by E. coli beta-glucuronidase using the on-line benzodiazepine screening immunoassay on the Roche/Hitachi 917 analyzer.
Carol L O'Neal et al.
Journal of forensic sciences, 47(2), 427-428 (2002-03-23)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service