Skip to Content
Merck
All Photos(1)

Key Documents

N-902

Supelco

Nortriptyline-D3 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19D3H18N · HCl
CAS Number:
Molecular Weight:
302.86
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])NCCC=C1C2=C(C=CC=C2)CCC3=C1C=CC=C3.[H]Cl

InChI

1S/C19H21N.ClH/c1-20-14-6-11-19-17-9-4-2-7-15(17)12-13-16-8-3-5-10-18(16)19;/h2-5,7-11,20H,6,12-14H2,1H3;1H/i1D3;

InChI key

SHAYBENGXDALFF-NIIDSAIPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nortriptyline is a second generation tricyclic antidepressant (TCA) marketed under trade names such as Sensoval, Aventyl®, and Pamelor® for treatment of major depression. Nortriptyline is also an active metabolite of the TCA amitriptyline. This certified Snap-N-Spike® Solution is suitable as an internal standard for quantitation of nortriptyline levels by LC/MS or GC/MS in clinical toxicology, forensic analysis, urine drug testing, isotope dilution methods, or medication monitoring applications.

Application


  • Quantification of Tricyclic Antidepressants in Serum Using Liquid Chromatography Electrospray Tandem Mass Spectrometry (HPLC-ESI-MS/MS).: This study demonstrates a robust method for the quantification of tricyclic antidepressants, including Nortriptyline-D3, using high-performance liquid chromatography coupled with electrospray ionization tandem mass spectrometry. The approach offers precise measurement essential for pharmacokinetic and neuropharmacological research, aligning with the interests of researchers in pharma and biotech focused on antidepressant metabolism studies (Clarke et al., 2016).


Legal Information

Aventyl is a registered trademark of Eli Lilly and Co.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Pamelor is a registered trademark of Mallinckrodt Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service