Skip to Content
Merck
All Photos(1)

Key Documents

M-165

Supelco

5-MeO-DALT solution

1.0 mg/mL in acetonitrile, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22N2O
CAS Number:
Molecular Weight:
270.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

format

single component solution

storage temp.

−20°C

SMILES string

C=CCN(CC=C)CCC1=CNC2=CC=C(OC)C=C21

InChI

1S/C17H22N2O/c1-4-9-19(10-5-2)11-8-14-13-18-17-7-6-15(20-3)12-16(14)17/h4-7,12-13,18H,1-2,8-11H2,3H3

InChI key

HGRHWEAUHXYNNP-UHFFFAOYSA-N

General description

Classified as a novel psychoactive substance (NPS) by the United Nations Office on Drugs and Crime (UNODC), 5-MeO-DALT is a hallucinogenic tryptamine that has reports of abuse in Europe and in the United States. This certified Snap-N-Spike® solution is suitable for a variety of GC/MS or LC/MS testing applications including for use in forensic analysis, urine drug testing, or clinical toxicology.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service