Skip to Content
Merck
All Photos(1)

Key Documents

M-108

Supelco

MTBSTFA

with 1% t-BDMCS, ampule of 5 × 1 mL, analytical standard, Cerilliant®

Synonym(s):

N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-Butyldimethylchlorosilane, N-Methyl-N-tert-butyldimethylsilyltrifluoroacetamide, MTBSTFA + 1% TBDMSCl, Silylating mixture VI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18F3NOSi
CAS Number:
Molecular Weight:
241.33
Beilstein:
3606546
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

product name

MTBSTFA (with 1% t-BDMCS), ampule of 5 × 1 mL, (with 1% t-BDMCS), analytical standard, Cerilliant®

grade

analytical standard

Quality Level

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

packaging

ampule of 5 × 1 mL

manufacturer/tradename

Cerilliant®

technique(s)

gas chromatography (GC): suitable

application(s)

forensics and toxicology

format

neat

storage temp.

−20°C

SMILES string

CN(C(=O)C(F)(F)F)[Si](C)(C)C(C)(C)C

InChI

1S/C9H18F3NOSi/c1-8(2,3)15(5,6)13(4)7(14)9(10,11)12/h1-6H3

InChI key

QRKUHYFDBWGLHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Pharmaceutical substances in ambient particulates: A preliminary assessment.: This study explores the presence of pharmaceutical compounds in environmental particulate matter, using analytical methods that could involve derivatization agents like MTBSTFA. The detection of these substances in air samples suggests potential health impacts and environmental persistence (Cecinato et al., 2017) (Cecinato et al., 2017).


Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ho-Sang Shin et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(7-8), 489-494 (2011-02-01)
A gas chromatography-mass spectrometric (GC-MS) method was developed for the determination of 2-naphthol (2-NAP) and 1-hydroxypyrene (1-HOP) in human urine. Extraction from urine after the enzyme hydrolysis with β-glucuronidase/arylsulfatase was achieved with a liquid extraction using 5 mL of pentane.
Xiaowei Wang et al.
Journal of chromatography. A, 1216(35), 6267-6273 (2009-07-31)
A sample pretreatment method for the determination of 18 chlorophenols (CPs) in aqueous samples by derivatization liquid-phase microextraction (LPME) was investigated using gas chromatography-mass spectrometry. Derivatization reagent was spiked into the extraction solvent to combine derivatization and extraction into one
Martin Adiels et al.
Rapid communications in mass spectrometry : RCM, 24(5), 586-592 (2010-02-09)
Stable isotope kinetic studies play an important role in the study of very-low density lipoprotein (VLDL) metabolism, including basic and clinical research. Today, [1,1,2,3,3-(2)H(5)]glycerol is the most cost-effective alternative to measure glycerol and triglyceride kinetics. Recycling of glycerol from glycolysis
R Montes et al.
Journal of chromatography. A, 1216(2), 205-210 (2008-12-17)
A fast and novel sample preparation procedure for the determination of triclosan (TCS) and methyltriclosan (MTCS) in water samples is presented. Dispersive liquid-liquid microextraction, using a ternary mixture consisting of a disperser, an extractant and N-methyl-N-(tert-butyldimethylsilyl)trifluoroacetamide (MTBSTFA) as derivatization reagent
P Canosa et al.
Journal of chromatography. A, 1072(1), 107-115 (2005-05-11)
A solid-phase microextraction (SPME) method for the determination of triclosan, methyl triclosan, 2,4-dichlorophenol and 2,3,4-trichlorophenol (considered as possible triclosan metabolites) in water samples was optimised. Analytes were first concentrated on a SPME fibre, directly exposed to the sample, and then

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service