Skip to Content
Merck
All Photos(1)

Key Documents

H-085

Supelco

17α-Hydroxyprogesterone solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O3
CAS Number:
Molecular Weight:
330.46
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C21H30O3/c1-13(22)21(24)11-8-18-16-5-4-14-12-15(23)6-9-19(14,2)17(16)7-10-20(18,21)3/h12,16-18,24H,4-11H2,1-3H3/t16-,17+,18+,19+,20+,21+/m1/s1

InChI key

DBPWSSGDRRHUNT-CEGNMAFCSA-N

General description

A calibration material for use in newborn screening assays by LC-MS/MS. This Certified Spiking Solution® is suitable for many critical applications in clinical and research environments from preparation of controls and system tuning/suitability standards to OEM use in preparation of reagents and assay development.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hye Jeong Kim et al.
Journal of Korean medical science, 27(11), 1439-1443 (2012-11-21)
Congenital adrenal hyperplasia (CAH) is characterized by decreased adrenal hormone production due to enzymatic defects and subsequent rise of adrenocorticotrophic hormone that stimulates the adrenal cortex to become hyperplastic, and sometimes tumorous. As the pathophysiology is basically a defect in
O Lekarev et al.
Journal of perinatology : official journal of the California Perinatal Association, 33(1), 76-78 (2012-12-28)
Congenital adrenal hyperplasia (CAH) is an autosomal recessive disorder where steroidogenesis in the adrenal cortex is impaired. The most common form is caused by 21-hydroxylase deficiency (21OHD). Classical 21OHD is characterized by glucocorticoid and mineralocorticoid deficiency and by overproduction of
Adriana Pabón et al.
Molecules (Basel, Switzerland), 18(3), 3356-3378 (2013-03-16)
Solanum nudum Dunal steroids have been reported as being antimalarial compounds; however, their concentration in plants is low, meaning that the species could be threatened by over-harvesting for this purpose. Swern oxidation was used for hemisynthesis of diosgenone (one of
Shilpi Singh et al.
Diabetes & metabolic syndrome, 6(4), 207-211 (2012-12-04)
An interaction between adiponectin, steroid synthesis or action and measures of insulin resistance (IR) have been reported in the pathogenesis of polycystic ovary syndrome (PCOS). The present study was done to determine plasma adiponectin concentration (PAC) in women with and
K Deveci et al.
European review for medical and pharmacological sciences, 17(2), 261-265 (2013-02-05)
Familial Mediterranean Fever (FMF) is an autosomal recessive disease characterized by short lived, febrile serosae inflammatory attacks. FMF has various effects in multiple systems and organs. In the present study, our aim was to evaluate adrenal steroidogenesis in female FMF

Articles

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service