Key Documents
Z743191
Julabo® bath test tube racks
to fit 21 tubes, diam. 30 mm, plastic
Synonym(s):
tube holder
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
material
plastic
packaging
pack of 1 ea
manufacturer/tradename
Julabo 9970303
parameter
100 °C max. temp.
diam.
30 mm
General description
Plastic test tube insert holds 21 test tubes, 30 mm diameter, can be used at temperatures up to 100°C. When tempering samples containing solvents or acids above 60°C, it is recommended using stainless steel test glass inserts, as the plastic may be attacked.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.