Skip to Content
Merck
All Photos(1)

Key Documents

Z742928

Detonox® detergent, ultimate precision cleaner

pkg of 3.8 L

Synonym(s):

USDA authorized cleaner, alkaline detergent, emulsifying detergent, low foam cleaner, machine washing detergent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800
NACRES:
NB.34

packaging

pkg of 3.8 L
pkg of 4 × 3.8 L

manufacturer/tradename

Alconox, Inc. 2301 (4 x 3.8 L)
Alconox, Inc. 2301-1 (3.8 L)

General description

Detonox Ultimate Precision Cleaner

  • Concentrated to save you money
  • Exceptional cosmetic and pharmaceutical polymer, cream and lotion removal
  • Non-caustic for greater worker safety and greater material compatibility
  • Free rinsing to give you reliable results and no interfering residues

Used to remove: Cosmetic bases and formulations, organics, grease, tars, resins, waxes, fats, oils, titanium and zinc oxides, ointments and lotions.

Surfaces cleaned: Recommended for glass, stainless steel, PTFE, ceramic, cement and solvent resistant plastics and metals. Corrosion testing is advisable.

Cleaning method: Soak, circulate under immersions, ultrasonic cleaners, and manual cleaning.

Diections:
Make a fresh 1-3% solution (1 ¼ - 3 3/4 oz. per gal. or 10-30 ml per liter) in cold, warm or hot water. If available, use very hot water to clean cosmetic residues. For difficult soils, raise water temperature and use more detergent. Elevation of temperature is true under most circumstances. Check with Alconox Inc, for use of hot water if products contain Titanium Oxide (TiO) or Zinc Oxide (ZnO) as
they are notable exceptions to heat expediting detergency. Clean by soak, circulate, wipe or ultrasonic method. Not for spray machines, will foam.
RINSE THOROUGHLY—preferably with running water. For critical cleaning, do final or all rinsing in purified water.

Caution: Contains anionic and nonionic surfactants with phosphate chelating agents. May cause skin and eye irritation. Wear eye protection and chemical resistant gloves.

Legal Information

Detonox is a registered trademark of Alconox, Inc.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service