Key Documents
Z684457
Sigma-Aldrich
Aldrich® solvent storage/dispensing flask, septum-inlet, with PTFE inlet valve
joint: ST/NS 29/32, capacity 100 mL
Synonym(s):
solvent dispenser bottles
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
glass
round bottom flask
single-neck
Joint
joint: ST/NS29/32
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 1
manufacturer/tradename
Sigma-Aldrich
capacity
100 mL
General description
Designed for use with PureSolv Micro systems with a standard taper dispensing joint. PTFE inlet valve may be closed after filling to isolate dry solvent and to remove flask from system and take to another location for use. Use septum Z565695 or Z565679.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC