Skip to Content
Merck
All Photos(1)

Key Documents

W313726

Sigma-Aldrich

2,6-Dimethoxyphenol

natural, ≥96%

Synonym(s):

Pyrogallol 1,3-dimethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3OH
CAS Number:
Molecular Weight:
154.16
FEMA Number:
3137
Beilstein:
1526871
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.036
NACRES:
NA.21

grade

Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

Assay

≥96%

bp

261 °C (lit.)

mp

50-57 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

bacon; balsamic; smoky

SMILES string

COc1cccc(OC)c1O

InChI

1S/C8H10O3/c1-10-6-4-3-5-7(11-2)8(6)9/h3-5,9H,1-2H3

InChI key

KLIDCXVFHGNTTM-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

General description

2,6-Dimethoxyphenol (syringol) is a phenol with methoxy groups at the 2 and 6 positions. It is a highly versatile molecule with a woody, and smoky aroma used in foods like meat, soups, and seafood.

Application

2,6-Dimethoxyphenol is used as a flavoring agent in the food industry, particularly in smoked foods.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

284.0 °F - closed cup

Flash Point(C)

140 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seperation of syringol from birch wood-derived vacuum pyrolysis oil
Murwanashyaka JN, et al.
Separation and Purification Technology, 24(1-2), 155-165 (2001)
Chemical composition, antioxidant, and antibacterial activity of wood vinegar from Litchi chinensis
Yang JF, et al.
Molecules (Basel), 21(9), 1150-1150 (2016)
Joseph Zakzeski et al.
ChemSusChem, 4(3), 369-378 (2011-01-20)
The solubilization and aqueous phase reforming of lignin, including kraft, soda, and alcell lignin along with sugarcane bagasse, at low temperatures (T≤498 K) and pressures (P≤29 bar) is reported for the first time for the production of aromatic chemicals and hydrogen. Analysis
Isabel Pardo et al.
Biotechnology and bioengineering, 109(12), 2978-2986 (2012-06-26)
DNA recombination methods are useful tools to generate diversity in directed evolution protein engineering studies. We have designed an array of chimeric laccases with high-redox potential by in vitro and in vivo DNA recombination of two fungal laccases (from Pycnoporus
R Taboada-Puig et al.
Bioresource technology, 102(11), 6593-6599 (2011-04-21)
Versatile peroxidase (VP) from Bjerkandera adusta was insolubilized in the form of cross-linked enzyme aggregates (CLEA®s). Of the initially applied activity 67% was recovered as CLEA®s. Co-aggregation of VP with glucose oxidase from Aspergillus niger led to an increased activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service