Skip to Content
Merck
All Photos(4)

Key Documents

W270215

Sigma-Aldrich

5-Methylfurfural

greener alternative

mixture of isomers, ≥99%, natural, FG

Synonym(s):

2-Furancarboxaldehyde 5-methyl, 5-Methyl-2-furaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O2
CAS Number:
Molecular Weight:
110.11
FEMA Number:
2702
Beilstein:
106895
EC Number:
MDL number:
UNSPSC Code:
12164502
Flavis number:
13.001
NACRES:
NA.21

biological source

botanical

Quality Level

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.510

Assay

≥99%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.531 (lit.)

bp

187 °C (lit.)

density

1.107 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

bread; caramel; brown; spicy; sweet

storage temp.

2-8°C

SMILES string

[H]C(=O)c1ccc(C)o1

InChI

1S/C6H6O2/c1-5-2-3-6(4-7)8-5/h2-4H,1H3

InChI key

OUDFNZMQXZILJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is Biobased and thus aligns with "Less Hazardous Chemical Syntheses" and "Use of Renewable Feedstocks".

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service