Skip to Content
Merck
All Photos(1)

Key Documents

W261505

Sigma-Aldrich

Lauric aldehyde

≥95%, stabilized, FCC, FG

Synonym(s):

Dodecyl aldehyde, Aldehyde C12, Dodecanal, Lauraldehyde, Laurinaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CHO
CAS Number:
Molecular Weight:
184.32
FEMA Number:
2615
Beilstein:
1703917
EC Number:
Council of Europe no.:
99
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.011
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥95%

contains

α-tocopherol as stabilizer

refractive index

n20/D 1.435 (lit.)

bp

185 °C/100 mmHg (lit.)

density

0.831 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; citrus; waxy; floral; soapy

SMILES string

CCCCCCCCCCCC=O

InChI

1S/C12H24O/c1-2-3-4-5-6-7-8-9-10-11-12-13/h12H,2-11H2,1H3

InChI key

HFJRKMMYBMWEAD-UHFFFAOYSA-N

General description

Lauric aldehyde is one of the main aliphatic green volatile compounds in macerated grape cluster stems. It is also one of the key flavor constituents of grapefruit oil.

Application


  • Utility of ToxTracker in animal alternative testing strategy for fragrance materials.: This study highlights the use of ToxTracker, a suite of reporter cell lines, to evaluate the genetic toxicity of fragrance materials, including lauric aldehyde, offering a potential alternative to animal testing and advancing the safety assessment of consumer products (Thakkar et al., 2023).

  • In vitro evaluation of polymeric micelles based on hydrophobically-modified sulfated chitosan as a carrier of doxorubicin.: Research demonstrates the potential of hydrophobically-modified sulfated chitosan micelles, incorporating lauric aldehyde, for improving the delivery of chemotherapeutic agents, particularly doxorubicin, enhancing drug stability and reducing side effects (Wang et al., 2012).

  • Biocompatibility of injectable chitosan-phospholipid implant systems.: Investigates the biocompatibility of chitosan-phospholipid injectable implants, where lauric aldehyde could play a role in modifying polymer matrices to enhance biocompatibility and drug release profiles, critical for medical implant technologies (De Souza et al., 2009).


Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Green odorants of grape cluster stem and their ability to cause a wine stemmy flavor.
Hashizume K & Samuta T.
Journal of Agricultural and Food Chemistry, 45(4), 1333-1337 (1997)
Analysis of carbonyl flavor constituents from grapefruit oil.
Moshonas MG.
Journal of Agricultural and Food Chemistry, 19(4), 769-770 (1971)
Markus A Keller et al.
Nature communications, 5, 4439-4439 (2014-07-23)
Mutations in the gene coding for membrane-bound fatty aldehyde dehydrogenase (FALDH) lead to toxic accumulation of lipid species and development of the Sjögren-Larsson Syndrome (SLS), a rare disorder characterized by skin defects and mental retardation. Here, we present the crystallographic
James Liu et al.
Journal of pharmaceutical sciences, 104(3), 1187-1196 (2015-01-13)
The objective of this study is to develop and compare several Sorafenib-loaded biocompatible nanoparticle models in order to optimize drug delivery and tumor cellular kill thereby improving the quality of Sorafenib-regimented chemotherapy. Sorafenib-loaded poly (lactic-co-glycolic) acid (PLGA), 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC) liposomes
Pingxi Xu et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(46), 16592-16597 (2014-10-29)
Insect repellents are important prophylactic tools for travelers and populations living in endemic areas of malaria, dengue, encephalitis, and other vector-borne diseases. DEET (N,N-diethyl-3-methylbenzamide) is a 6-decade-old synthetic repellent, which is still considered the gold standard of mosquito repellents. Mosquitoes

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service