Skip to Content
Merck
All Photos(1)

Key Documents

W237604

Sigma-Aldrich

Diethyl sebacate

≥98%, FCC, FG

Synonym(s):

Diethyl decanedioate, Decanedioic acid 1,10-diethyl ester, NSC 8911, Sebacic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCO(CH2)8COOC2H5
CAS Number:
Molecular Weight:
258.35
FEMA Number:
2376
Beilstein:
1790779
EC Number:
Council of Europe no.:
623
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.475
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.436 (lit.)

bp

312 °C (lit.)

mp

1-2 °C (lit.)

density

0.963 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

melon; quince; wine-like

SMILES string

CCOC(=O)CCCCCCCCC(=O)OCC

InChI

1S/C14H26O4/c1-3-17-13(15)11-9-7-5-6-8-10-12-14(16)18-4-2/h3-12H2,1-2H3

InChI key

ONKUXPIBXRRIDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl sebacate is a volatile organic compoind used as a flavoring agent. It is reported to be found in longan honey porduced by Apis cerana.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burdock, GA
Encyclopedia of Food and Color Additives, 1, 821-822 (1997)
Volatile organic compounds of Thai honeys produced from several floral sources by different honey bee species
Pattamayutanon P, et al.
PLoS ONE, 12(2), e0172099-e0172099 (2017)
Burdock, GA
Encyclopedia of Food and Color Additives, 1, 821-822 (1997)
Y Shoji et al.
The Journal of pharmacy and pharmacology, 46(8), 643-646 (1994-08-01)
The effects of a topical preparation of mycophenolic acid on the experimental allergic contact dermatitis induced by dinitrofluorobenzene was investigated. Visual assessment of skin reactions showed significant efficacy of a topical preparation of mycophenolic acid. This efficacy appeared from the
Allergic contact dermatitis from diethyl sebacate in lanoconazole cream.
M Tanaka et al.
Contact dermatitis, 43(4), 233-234 (2000-09-30)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service