Skip to Content
Merck
All Photos(1)

Key Documents

V1309

Sigma-Aldrich

3,4-Dimethoxybenzylamine

97%

Synonym(s):

Veratrylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3,4-(CH3O)2C6H3CH2NH2
CAS Number:
Molecular Weight:
167.21
Beilstein:
511575
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.556 (lit.)

bp

281-284 °C (lit.)

density

1.109 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(CN)cc1OC

InChI

1S/C9H13NO2/c1-11-8-4-3-7(6-10)5-9(8)12-2/h3-5H,6,10H2,1-2H3

InChI key

DIVNUTGTTIRPQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

<ul>
<li><strong>Fluorescence Derivatization for Serotonin Determination:</strong> 3,4-Dimethoxybenzylamine is employed as a sensitive pre-column fluorescence derivatization reagent for measuring serotonin levels in human platelet-poor plasma, enhancing detection sensitivity and specificity (Ishida et al., 1997).</li>
<li><strong>Spectrofluorimetric Analysis of 5-Hydroxyindoles:</strong> This compound serves as a selective fluorogenic reagent for the spectrofluorimetric determination of 5-hydroxyindoles, contributing to accurate and selective analysis techniques in clinical and biochemical studies (Ishida et al., 1991).</li>
</ul>

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Ishida et al.
The Analyst, 116(3), 301-304 (1991-03-01)
A spectrofluorimetric method has been developed for the sensitive and selective determination of 5-hydroxyindoles; the method is based on the reaction of 5-hydroxyindoles in a weakly alkaline solution (pH 9.0) with aromatic methylamines in the presence of potassium hexacyanoferrate(III) and
J Ishida et al.
Journal of chromatography. B, Biomedical sciences and applications, 692(1), 31-36 (1997-04-25)
3,4-Dimethoxybenzylamine is shown to be a highly sensitive pre-column fluorescence derivatization reagent for the determination of serotonin in plasma by high-performance liquid chromatography. The reagent reacts selectively with 5-hydroxyindoles including serotonin in slightly alkaline media in the presence of potassium
A M Feigin et al.
Neuroreport, 6(16), 2134-2136 (1995-11-13)
The irritating, pungent compound, capsaicin (10-20 microM), induces the formation of non-selective ion channels with a wide variety of conductances in protein-free lipid bilayers form from a mixture of zwitterionic phospholipids. The channel-forming activity of capsaicin and four of its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service