Skip to Content
Merck
All Photos(2)

Key Documents

T7633

Sigma-Aldrich

Triethylenetetramine-N,N,N′,N′′,N′′′,N′′′-hexaacetic acid

for complexometry, ≥98.0%

Synonym(s):

TTHA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2N(CH2CO2H)CH2CH2N(CH2CO2H)2]2
CAS Number:
Molecular Weight:
494.45
Beilstein:
1811090
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

for complexometry

Assay

≥98.0%

form

powder

mp

232-233 °C (lit.)

SMILES string

OC(=O)CN(CCN(CCN(CC(O)=O)CC(O)=O)CC(O)=O)CCN(CC(O)=O)CC(O)=O

InChI

1S/C18H30N4O12/c23-13(24)7-19(3-5-21(9-15(27)28)10-16(29)30)1-2-20(8-14(25)26)4-6-22(11-17(31)32)12-18(33)34/h1-12H2,(H,23,24)(H,25,26)(H,27,28)(H,29,30)(H,31,32)(H,33,34)

InChI key

RAEOEMDZDMCHJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agnieszka Gawin et al.
Scientific reports, 10(1), 12470-12470 (2020-07-29)
Bacterial alginate initially consists of 1-4-linked β-D-mannuronic acid residues (M) which can be later epimerized to α-L-guluronic acid (G). The family of AlgE mannuronan C-5-epimerases from Azotobacter vinelandii has been extensively studied, and three genes putatively encoding AlgE-type epimerases have
S M Eleff et al.
Magnetic resonance in medicine, 30(1), 11-17 (1993-07-01)
The intracellular to extracellular sodium distribution is one of the primary determinants of action potentials necessary for the electrical function of organs such as brain, heart and skeletal muscle. The ability of shift reagent enhanced 23Na MRS to directly measure
Y Boulanger et al.
NMR in biomedicine, 5(1), 1-10 (1992-01-01)
The effects of two widely used paramagnetic shift reagents for cationic NMR, dysprosium tripolyphosphate [Dy(PPP)2(7-)] and dysprosium triethylenetetramine hexaacetate [Dy(TTHA)3-], on the cell structure of dog and human erythrocytes, dog kidney cortical tubules and rat hepatocytes were investigated. The effect
Lev N Krasnoperov et al.
Bioconjugate chemistry, 21(2), 319-327 (2010-01-21)
Novel amino-reactive derivatives of lanthanide-based luminescent labels of enhanced brightness and metal retention were synthesized and used for the detection of cDNA oligonucleotides by molecular beacons. Time-resolved acquisition of the luminescent signal that occurs upon hybridization of the probe to
Concetta De Stefano et al.
Analytical and bioanalytical chemistry, 375(7), 956-967 (2003-04-23)
The binding ability of diethylene triamine pentaacetate (dtpa(5-)) and triethylene tetraamine hexaacetate (ttha(6-)) ligands towards major components, H(+), Na(+), Mg(2+) and Ca(2+), of natural waters was studied in both single and mixed ionic media at different ionic strengths and at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service