Skip to Content
Merck
All Photos(1)

Key Documents

T46051

Sigma-Aldrich

s-Triazine

97%

Synonym(s):

1,3,5-Triazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H3N3
CAS Number:
Molecular Weight:
81.08
Beilstein:
104790
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

77-83 °C (dec.) (lit.)

SMILES string

c1ncncn1

InChI

1S/C3H3N3/c1-4-2-6-3-5-1/h1-3H

InChI key

JIHQDMXYYFUGFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nazli Alizadeh-Tabrizi et al.
Environmental toxicology and pharmacology, 49, 8-13 (2016-11-25)
The current study designed to clarify the mechanism of paraquat-induced cytotoxicity and protective effects of Atorvastatin on freshly isolated alveolar macrophages (AMs). AMs were collected via bronchoalveolar lavage and exposed to various concentrations of paraquat in the presence and absence
Amit Kumar et al.
Environmental pollution (Barking, Essex : 1987), 259, 113765-113765 (2019-12-31)
We utilized volcanic CO2 vents at Castello Aragonese off Ischia Island as a natural laboratory to investigate the effect of lowered pH/elevated CO2 on the bioactivities of extracts from fleshy brown algae Sargassum vulgare C. Agardh. We analysed the carbohydrate
Qing-Ping Hu et al.
Scientific reports, 7, 45231-45231 (2017-03-25)
Cyperus rotundus L. (Cyperaceae) is a medicinal herb traditionally used to treat various clinical conditions at home. In this study, chemical composition of Cyperus rotundus rhizomes essential oil, and in vitro antioxidant, DNA damage protective and cytotoxic activities as well
Jerzy Drzewiecki et al.
Applied biochemistry and biotechnology (2018-05-10)
Electrophoretic, antioxidant, and FTIR profiles of some varieties of amaranth, quinoa, and buckwheat seeds and their by products were compared. Water extracts of these products were evaluated by the Folin-Ciocalteau method in order to determine total phenolic content. The antioxidant
Ruixia Lan et al.
Animals : an open access journal from MDPI, 10(1) (2020-01-01)
Oxidative stress is induced by excessive oxidative radicals, which directly react with biomolecules, and damage lipids, proteins and DNA, leading to cell or organ injury. Supplementation of antioxidants to animals can be an effective way to modulate the antioxidant system.

Related Content

As the exploration of the properties of complex natural products becomes increasingly more sophisticated with the technological advances being made in their screening and evaluation and as structural details of their interaction with biological targets becomes more accessible, the importance and opportunities for providing unique solutions to complex biological problems has grown. The Boger Lab addresses these challenging problems by understanding the complex solutions and subtle design elements that nature has provided in the form of a natural product and work to extend the solution through rational design elements to provide more selective, more efficacious, or more potent agents designed specifically for the problem or target under investigation. The resulting efforts have reduced many difficult or intractable synthetic challenges to manageable problems providing an approach not only to the natural product but one capable of simple extrapolation to a series of structural analogs with improved selectivity and efficacy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service