Skip to Content
Merck
All Photos(1)

Key Documents

SIK7911-10

Sigma-Aldrich

[1,1′-Biphenyl]-4,4′-diylbis(phosphonic acid)

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12O6P2
CAS Number:
Molecular Weight:
314.17
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.23

Assay

97%

form

powder

shelf life

3 years under the recommended conditions

color

white

InChI

1S/C12H12O6P2/c13-19(14,15)11-5-1-9(2-6-11)10-3-7-12(8-4-10)20(16,17)18/h1-8H,(H2,13,14,15)(H2,16,17,18)

InChI key

QWSZMKCGMDROKE-UHFFFAOYSA-N

Application

These products are used as linkers to improve adhesion between coatings and metal surfaces. The phosphonic acid groups can graft onto metal surface while the hydroxyl groups react with common epoxy/amine-based paints.

Legal Information

Product of SiKEMIA

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service