Skip to Content
Merck
All Photos(1)

Key Documents

M73788

Sigma-Aldrich

N-Methyl-2-piperidone

99%

Synonym(s):

1-Methyl-2-piperidone, 1-Methyl-6-oxopiperidine, N-Methyl-δ-valerolactam, N-Methyl-2-piperidinone, N-Methyl-2-piperidone, N-Methylpiperidone, N-Methylvalerolactam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO
CAS Number:
Molecular Weight:
113.16
Beilstein:
1708
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

bp

105-106 °C/12 mmHg (lit.)

density

0.997 g/mL at 25 °C (lit.)

SMILES string

CN1CCCCC1=O

InChI

1S/C6H11NO/c1-7-5-3-2-4-6(7)8/h2-5H2,1H3

InChI key

GGYVTHJIUNGKFZ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ekaterina F Kolesanova et al.
International journal of peptides, 2013, 197317-197317 (2013-09-13)
Unified schedule for multiple parallel solid-phase synthesis of so-called "difficult" peptides on polypropylene pins was developed. Increase in the efficiency of 9-fluorenyl(methoxycarbonyl) N-terminal amino-protecting group removal was shown to have a greater influence on the accuracy of the "difficult" peptide
Ilnaz T Rakipov et al.
Molecules (Basel, Switzerland), 26(5) (2021-04-04)
In the present work, the thermochemistry of solution, solvation, and hydrogen bonding of cyclic amides in proton acceptor (B) and proton donor (RXH) solvents were studied. The infinite dilution solution enthalpies of δ-valerolactam, N-methylvalerolactam, ε-caprolactam, and N-methylcaprolactam were measured at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service