Skip to Content
Merck
All Photos(1)

Key Documents

H146

Sigma-Aldrich

4-Hydroxymephenytoin

≥98% (HPLC)

Synonym(s):

(±)-5-Ethyl-5-(4-hydroxyphenyl)-3-methylhydantoin, (±)-5-Ethyl-5-(4-hydroxyphenyl)-3-methylimididazolidine-2,4-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N2O3
CAS Number:
Molecular Weight:
234.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98% (HPLC)

solubility

DMSO: >5 mg/mL

SMILES string

CCC1(NC(=O)N(C)C1=O)c2ccc(O)cc2

InChI

1S/C12H14N2O3/c1-3-12(8-4-6-9(15)7-5-8)10(16)14(2)11(17)13-12/h4-7,15H,3H2,1-2H3,(H,13,17)

InChI key

OQPLORUDZLXXPD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

CYP2C19 metabolite of mephenytoin.

Application

Pharmacological activity investigating:
  • The drug effects on metabolization
  • In vitro cytochrome P450 activity in microsomes

Analyte for electron-capture gas chromatography procedures

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Salsali, et al.,
Journal of Pharmacological and Toxicological Methods, 44, 461-465 (2001)
Sonja Krösser et al.
European journal of clinical pharmacology, 62(4), 277-284 (2006-03-10)
The 5HT(1A) receptor agonist sarizotan is in clinical development for the treatment of dyskinesia, a potentially disabling complication in Parkinson's disease. We investigated the effect of sarizotan on the clinical pharmacokinetics of probe drugs for cytochrome P450 (CYP) to evaluate
Xiao-Ping Chen et al.
Clinical pharmacology and therapeutics, 73(3), 264-271 (2003-03-07)
This study was designed to define the effect of low-dose aspirin administration on the activity of cytochrome P450 (CYP) in normal human subjects. Aspirin, 50 mg daily, was given for 14 days to 18 nonsmoking healthy male volunteers. A modified
J Caslavska et al.
Journal of chromatography. B, Biomedical applications, 656(1), 219-231 (1994-06-03)
Phenotypes for hydroxylation can be predicted by using mephenytoin and dextromethorphan as substrates, whereas phenotypes for acetylation can be determined with caffeine as probe drug. After single-dose administration of one of these drugs, of two of them simultaneously, or of
Xunge Zhang et al.
Pharmaceutical biology, 58(1), 247-252 (2020-04-01)
Context: Cepharanthine (CEP) extracted from the roots of Stephania cepharantha Hayata (Menispermaceae), has a range of therapeutic potential in clinical conditions. Whether it affects the activity of human liver cytochrome P450 (CYP) enzymes remains unclear.Materials and methods: The effects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service