Skip to Content
Merck
All Photos(1)

Key Documents

D80800

Sigma-Aldrich

N,N′-Dicyclohexylurea

98%

Synonym(s):

1,3-Dicyclohexylurea

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11NHCONHC6H11
CAS Number:
Molecular Weight:
224.34
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

232-233 °C (lit.)

SMILES string

O=C(NC1CCCCC1)NC2CCCCC2

InChI

1S/C13H24N2O/c16-13(14-11-7-3-1-4-8-11)15-12-9-5-2-6-10-12/h11-12H,1-10H2,(H2,14,15,16)

InChI key

ADFXKUOMJKEIND-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mozhgan Aghajanzadeh et al.
International journal of pharmaceutics, 586, 119546-119546 (2020-06-17)
In the present study, we successfully synthesized nanocarriers (NCs) based on Y-shaped miktoarm copolymers, Poly Ethylene Glycol-Lysine-(Poly Caprolactone)2 (PEG-Lys-PCL2), which were loaded by baicalein (B) through the nanoprecipitation process to assess their in-vitro and in-vivo properties. We applied various methods
Stevan Pecic et al.
Bioorganic & medicinal chemistry letters, 22(1), 601-605 (2011-11-15)
Inhibition of soluble epoxide hydrolase (sEH) has been proposed as a new pharmaceutical approach for treating hypertension and vascular inflammation. The most potent sEH inhibitors reported in literature to date are urea derivatives. However, these compounds have limited pharmacokinetic profiles.
Xiang Fang et al.
American journal of physiology. Heart and circulatory physiology, 290(1), H55-H63 (2005-08-23)
Epoxyeicosatrienoic acids (EETs), lipid mediators synthesized from arachidonic acid by cytochrome P-450 epoxygenases, are converted by soluble epoxide hydrolase (SEH) to the corresponding dihydroxyeicosatrienoic acids (DHETs). Originally considered as inactive degradation products of EETs, DHETs have biological activity in some
Z Yu et al.
Circulation research, 87(11), 992-998 (2000-11-25)
The cytochrome P450-derived epoxyeicosatrienoic acids (EETs) have potent effects on renal vascular reactivity and tubular sodium and water transport; however, the role of these eicosanoids in the pathogenesis of hypertension is controversial. The current study examined the hydrolysis of the
Julian Dominik Wolf et al.
Journal of colloid and interface science, 566, 234-241 (2020-02-02)
Lysine based cationic surfactants are well-tolerated tools for hydrophobic ion pairing (HIP) with DNA and its incorporation into lipophilic delivery systems. Di-Boc-lysine was esterified with 1-hexadecanol and the Boc-residues were cleaved off resulting in hexadecyl lysinate (HL). Subsequently, its Log

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service