Key Documents
BR781530
Brand®1.2 ML Tubes
individual caps, PE
Synonym(s):
Accessories, Caps, Sample Storage, Tube racks, Tubes
Select a Size
10 ML
$40.80
100 ML
$67.50
250 ML
$85.40
500 ML
$155.00
1 L
$190.00
2.5 L
$302.00
$40.80
Estimated to ship on15 de marzo de 2025
To order products, please contact your local dealer.
Select a Size
Change View
10 ML
$40.80
100 ML
$67.50
250 ML
$85.40
500 ML
$155.00
1 L
$190.00
2.5 L
$302.00
About This Item
UNSPSC Code:
41122804
NACRES:
NB.22
$40.80
Estimated to ship on15 de marzo de 2025
To order products, please contact your local dealer.
material
PE
sterility
non-sterile
feature
individual caps
packaging
pack of 960 pieces (bulk packed)
manufacturer/tradename
BRAND 781530
capacity
1.2 mL
Compare Similar Items
View Full Comparison
Show Differences
1 of 4
This Item
BR781535BR781520BR781525
feature
individual caps
feature
strip of 8 caps
feature
individual tubes
feature
strip of 8 tubes
material
PE
material
polyethylene (cap), polypropylene (tubes and rack)
material
polypropylene
material
polypropylene
packaging
pack of 960 pieces (bulk packed)
packaging
pack of 120
packaging
pack of 960 (bulk)
packaging
pack of 120
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
capacity
1.2 mL
capacity
1.2 mL
capacity
1.2 mL
capacity
1.2 mL
General description
PE. Caps are free from RNase, DNase and DNA and not autoclavable. Ideal for biological tests, such as PCR, cell uptake studies, RIA and EIA. Designed for storing and freezing down to -80 °C, and for transporting reagents and samples. Compatible with standard microplates and suitable for working with multichannel pipettes. (Size of tubes: outer Ø 8.8 mm, height 45 mm.)BIO-CERT® CERTIFIED QUALITY: Free of human DNA, RNase, DNase and Pyrogens
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)