Skip to Content
Merck
All Photos(1)

Key Documents

935964

Sigma-Aldrich

Sodium hexafluorophosphate solution

new
greener alternative

in ethylene carbonate and dimethylcarbonate, 1.0 M NaPF6 in EC/DMC=50/50 (v/v), battery grade

Synonym(s):

1.0 M NaPF6 in EC/DMC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352600

grade

battery grade

Quality Level

description

Application: Battery Manufacturing

form

solution

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

(NaPF6 0.95-1.10 M)

impurities

≤100 ppm Acid
≤100.0 ppm H2O

application(s)

battery manufacturing

greener alternative category

General description

Sodium hexafluorophosphate solution in ethylene carbonate and dimethylcarbonate, 1.0 M NaPF6 in EC/DMC=25/75 (w/w) is a solution of battery-grade NaPF6 dissolved in binary mixture of EC/DMC=50/50 (v/v). The product is a clear solution with strictly minimized impurities including low moisture, low trace metals, low chloride, and low sulfate.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

1.0 M NaPF6 in EC/DMC=50/50 (v/v), battery grade is a superior electrolyte, designed for high-performance sodium-ion batteries Ethylene carbonate (EC) is a popular choice of solvent because it provides a high-dielectric constant to dissolve the electrolytic salt and helps to form an ionically conductive, passivating solid electrolyte interphase (SEI) on the electrodes. The addition of dimethylcarbonate (DMC) lowers the blend’s viscosity and melting point, yielding an electrolyte with a good balance between desirable electrochemical properties, high dielectric constant, and low viscosity

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

77.0 °F

Flash Point(C)

25 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High Stability and Long Cycle Life of Rechargeable Sodium-Ion Battery Using Manganese Oxide Cathode: A Combined Density Functional Theory (DFT) and Experimental Study
Pandit B, et al.
ACS Applied Materials & Interfaces, 13, 11433?11441-11433?11441 (2021)
What differentiates the transport properties of lithium electrolyte in ethylene carbonate mixed with diethylcarbonate from those mixed with dimethylcarbonate?
Uchida S, et al.
Journal of Power Sources, 511, 230423-230423 (2021)
Electrolyte Solvation Structure Design for Sodium Ion Batteries
Tian Z, et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 9, e2201207-e2201207 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service