Skip to Content
Merck
All Photos(1)

Key Documents

914711

Sigma-Aldrich

8-Biotinylamido-3,6-dioxaoctanoic acid

≥95%

Synonym(s):

8-Biotinylamido-3,6-dioxaoctanoic acid, Acid-terminated biotin linker, Biotin-O2Oc-OH, Biotinyl-AEEA, Biotinyl-Ado-OH, Biotinylation reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H27N3O6S
CAS Number:
Molecular Weight:
389.47
UNSPSC Code:
12352106

Assay

≥95%

form

powder

storage temp.

−20°C

Application

8-Biotinylamido-3,6-dioxaoctanoic acid is a versatile biotinylated linker that can be incorporated into chemical tools via its terminal amino group. Labeling materials or proteins with biotin provides a means to enrich and capture targets from biological systems.

Automate your Biotin tagging with Synple Automated Synthesis Platform (SYNPLE-SC002)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan-Mei Tian et al.
Bioorganic & medicinal chemistry letters, 29(5), 707-712 (2019-01-24)
The orphan nuclear receptor Nur77 (also known as TR3 or nerve growth factor-induced clone B NGFI-B) functions as a nuclear transcription factor in the regulation of target gene expression and plays a critical role in the regulation of differentiation, proliferation
Ayaka Miyachi et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(22), 6644-6650 (2010-04-27)
Self-assembly of peptides and proteins is a key feature of biological functions. Short amphiphilic peptides designed with a beta-sheet structure can form sophisticated nanofiber structures, and the fibers are available as nanomaterials for arranging biomolecules. Peptide FI (H-PKFKIIEFEP-OH) self-assembles into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service