Skip to Content
Merck
All Photos(2)

Key Documents

913014

Sigma-Aldrich

[1,3-Bis(diphenylphosphino)propane]dibromonickel(II)

≥95%

Synonym(s):

Ni(dppp)Br2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H26Br2NiP2
CAS Number:
Molecular Weight:
630.94
UNSPSC Code:
12352101

Assay

≥95%

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

>300 °C

Application

[1,3-Bis(diphenylphosphino)propane]dibromonickel(II) is catalyst for:
  • Suzuki-Miyaura cross-coupling reaction of aryl halides with arylboronic acids
  • Solid state metathesis polycondensation
  • Polymerization of poly(3-alkoxythiophene)s
  • Double bond isomerization

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Felicia Weber et al.
Organic letters, 17(12), 2952-2955 (2015-05-30)
A highly reactive nickel catalyst comprising NiCl2(dppp) or NiCl2(dppe) with zinc powder, ZnI2 and Ph2PH, was applied in the isomerization of terminal alkenes to Z-2-alkenes. The double-bond geometry of the 2-alkene can be controlled via the reaction temperature to yield

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service