Skip to Content
Merck
All Photos(1)

Key Documents

907251

Sigma-Aldrich

H-L-Photo-lysine HCl

≥98%

Synonym(s):

(S)-2-Amino-6-((2-(3-methyl-3H-diazirin-3-yl)ethoxy)carbonylamino)hexanoic acid hydrochloride, (S)-2-Amino-6-((2-(3H-diazirin-3-yl)propoxy)carbonylamino)hexanoic acid hydrochloride, 3′-Azibutyl-N-carbamoyl-lysine hydrochloride, Abk, Diazirine amino acid, Photo-Lys, Photo-crosslinking amino acid, Photoprobe building block

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21ClN4O4
CAS Number:
Molecular Weight:
308.76
UNSPSC Code:
12352209

Assay

≥98%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

availability

available only in USA

application(s)

peptide synthesis

storage temp.

−20°C

InChI

1S/C11H20N4O4/c1-11(14-15-11)5-7-19-10(18)13-6-3-2-4-8(12)9(16)17/h8H,2-7,12H2,1H3,(H,13,18)(H,16,17)/t8-/m0/s1

InChI key

LUCMNTLJAFTFDU-QMMMGPOBSA-N

Application

H-L-Photo-lysine HCl is a diazirine-containing lysine amino acid and multifunctional probe building blocks. Its incorporation into peptides or small-molecule probes and tools allows for photoaffinity labeling of cellular targets and protein-protein interactions upon UV light (~360 nm) irradiation to form a covalent bond. This and other multifunctional probe building blocks will continue to accelerate drug discovery research for probing cellular mechanisms, target ID/validation, and understanding traditionally undruggable targets. An Fmoc-protected version is also available as 907383.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service