Skip to Content
Merck
All Photos(1)

Key Documents

900218

Sigma-Aldrich

CdSe/ZnS core-shell type quantum dots

stabilized with octadecylamine ligands, fluorescence λem 600 nm, 5 mg/mL in toluene

Synonym(s):

Fluorescent nanocrystals, QDs

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdSe/ZnS
UNSPSC Code:
26111700
NACRES:
NA.23

Quality Level

form

dispersion

concentration

5 mg/mL in toluene

λmax

585

fluorescence

λem 600 nm
λem 600 nm±10 nm, quantum yield >50%

Looking for similar products? Visit Product Comparison Guide

General description

Reactive group: Octadecylamine.

Application

Highly purified CdSe/ZnS quantum dots with an octadecylamine surface contain extremely low free organic ligands and impurities. They are well suited for display, solid state lighting and solar cell applications. They can be used as an electroluminescence emitter in quantum dot based LEDs or embedded in a polymer matrix as the backlight of an LCD display. CdSe/ZnS quantum dots in solid form can be dispersed in most non-polar organic solvents such as toluene, chloroform, and hexane.

Legal Information

Product of Ocean Nanotech, LLC.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3

Flash Point(F)

39.2 °F

Flash Point(C)

4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhanao Tan et al.
Nano letters, 7(12), 3803-3807 (2007-11-03)
We report a multilayer solution-processed blue light-emitting diode based on colloidal core/shell CdS/ZnS nanocrystal quantum dots (QDs). At a low-operating voltage of 5.5 V, the device emits spectrally pure blue radiation at 460 nm with a narrow full-width-at-half-maximum bandwidth of
Qingjiang Sun et al.
ACS nano, 3(3), 737-743 (2009-03-25)
Owing to their narrow bright emission band, broad size-tunable emission wavelength, superior photostability, and excellent flexible-substrate compatibility, light-emitting diodes based on quantum dots (QD-LEDs) are currently under intensive research and development for multiple consumer applications including flat-panel displays and flat
Bright, multicoloured light-emitting diodes based on quantum dots.
Sun Q, et al.
Nature Photonics, 1, 717-722 (2007)

Articles

Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.

Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.

Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.

Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service